Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 80582976-80583231 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C4orf22
Entrez Gene ID
255119
Gene Name
chromosome 4 open reading frame 22
Gene Aliases
-
UniGene
Hs.527104
Gene Chromosome Location
Chr.4: 80321265-80963756 on Build GRCh38
Transcripts
NM_001206997.1 NM_152770.2 XM_011531816.2 XM_017007972.1 XM_017007973.1 XM_017007974.1 XM_017007975.1 XM_017007976.1 XM_017007977.1 XM_017007978.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4395398 c.323G>A p.G108E Chr.4: 80583173-80583173 on Build GRCh38
7239714 c.270A>G p.L90L Chr.4: 80583120-80583120 on Build GRCh38
1633811 c.279T>C p.N93N Chr.4: 80583129-80583129 on Build GRCh38
3409567 c.246G>A p.T82T Chr.4: 80583096-80583096 on Build GRCh38
5122351 c.245C>T p.T82M Chr.4: 80583095-80583095 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769784138 Chr.4: 80583055-80583055 on Build GRCh38
rs1000657550 Chr.4: 80583162-80583162 on Build GRCh38
rs1486020 Chr.4: 80583138-80583138 on Build GRCh38
rs951869943 Chr.4: 80583045-80583045 on Build GRCh38
rs267600275 Chr.4: 80583124-80583124 on Build GRCh38
rs746117164 Chr.4: 80583059-80583079 on Build GRCh38
rs371121490 Chr.4: 80583142-80583142 on Build GRCh38
rs564286336 Chr.4: 80583196-80583196 on Build GRCh38
rs776030207 Chr.4: 80583166-80583166 on Build GRCh38
rs952472936 Chr.4: 80583091-80583091 on Build GRCh38
rs372776163 Chr.4: 80583067-80583067 on Build GRCh38
rs139510159 Chr.4: 80583120-80583120 on Build GRCh38
rs374358632 Chr.4: 80583057-80583057 on Build GRCh38
rs779046322 Chr.4: 80583048-80583057 on Build GRCh38
rs376455140 Chr.4: 80583041-80583041 on Build GRCh38
rs771283632 Chr.4: 80583077-80583077 on Build GRCh38
rs979925088 Chr.4: 80583018-80583018 on Build GRCh38
rs190071166 Chr.4: 80583068-80583068 on Build GRCh38
rs150179610 Chr.4: 80583167-80583167 on Build GRCh38
rs751823292 Chr.4: 80583109-80583109 on Build GRCh38
rs751700524 Chr.4: 80583172-80583172 on Build GRCh38
rs772318629 Chr.4: 80583064-80583064 on Build GRCh38
rs148301535 Chr.4: 80583034-80583034 on Build GRCh38
rs770100756 Chr.4: 80583161-80583161 on Build GRCh38
rs534105473 Chr.4: 80583075-80583075 on Build GRCh38
rs763099246 Chr.4: 80583099-80583099 on Build GRCh38
rs753522839 Chr.4: 80583116-80583116 on Build GRCh38
rs370746109 Chr.4: 80583072-80583072 on Build GRCh38
rs764254311 Chr.4: 80583169-80583169 on Build GRCh38
rs77136026 Chr.4: 80583182-80583182 on Build GRCh38
rs780064332 Chr.4: 80583123-80583123 on Build GRCh38
rs757609223 Chr.4: 80583111-80583111 on Build GRCh38
rs565037474 Chr.4: 80583064-80583064 on Build GRCh38
rs936084186 Chr.4: 80583190-80583190 on Build GRCh38
rs372340494 Chr.4: 80583033-80583033 on Build GRCh38
rs1046335998 Chr.4: 80583036-80583036 on Build GRCh38
rs745777979 Chr.4: 80583049-80583049 on Build GRCh38
rs759979773 Chr.4: 80583147-80583147 on Build GRCh38
rs765782959 Chr.4: 80583095-80583095 on Build GRCh38
rs377463947 Chr.4: 80583122-80583122 on Build GRCh38
rs750136970 Chr.4: 80583192-80583192 on Build GRCh38
rs200221458 Chr.4: 80583090-80583090 on Build GRCh38
rs749233735 Chr.4: 80583132-80583132 on Build GRCh38
rs963741195 Chr.4: 80583115-80583115 on Build GRCh38
rs78938046 Chr.4: 80583080-80583080 on Build GRCh38
rs767810443 Chr.4: 80583184-80583184 on Build GRCh38
rs146316801 Chr.4: 80583100-80583100 on Build GRCh38
rs142731425 Chr.4: 80583137-80583137 on Build GRCh38
rs141410009 Chr.4: 80583096-80583096 on Build GRCh38
rs1012110128 Chr.4: 80583016-80583016 on Build GRCh38
rs771728913 Chr.4: 80583145-80583145 on Build GRCh38

View Full Product Details