Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 70234711-70234984 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL9A1
Entrez Gene ID
1297
Gene Name
collagen type IX alpha 1
Gene Aliases
DJ149L1.1.2, EDM6, MED, STL4
UniGene
Hs.590892
Gene Chromosome Location
Chr.6: 70216040-70303083 on Build GRCh38
Transcripts
NM_001851.4 NM_078485.3 XM_011535429.2 XM_011535430.2 XM_017010246.1 XM_017010247.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3777813 c.1431G>A p.R477R Chr.6: 70234893-70234893 on Build GRCh38
3629328 c.2205G>A p.R735R Chr.6: 70234848-70234848 on Build GRCh38
6373161 c.1499C>T p.A500V Chr.6: 70234825-70234825 on Build GRCh38
3394293 c.1474C>T p.R492W Chr.6: 70234850-70234850 on Build GRCh38
3629335 c.2152G>A p.G718R Chr.6: 70234901-70234901 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs868065305 Chr.6: 70234900-70234900 on Build GRCh38
rs759552165 Chr.6: 70234747-70234747 on Build GRCh38
rs368781924 Chr.6: 70234810-70234810 on Build GRCh38
rs763477632 Chr.6: 70234783-70234783 on Build GRCh38
rs184749864 Chr.6: 70234952-70234952 on Build GRCh38
rs1014687667 Chr.6: 70234833-70234833 on Build GRCh38
rs192467838 Chr.6: 70234894-70234894 on Build GRCh38
rs762955706 Chr.6: 70234939-70234939 on Build GRCh38
rs764717285 Chr.6: 70234926-70234926 on Build GRCh38
rs149459564 Chr.6: 70234813-70234813 on Build GRCh38
rs887513791 Chr.6: 70234805-70234805 on Build GRCh38
rs749881260 Chr.6: 70234935-70234935 on Build GRCh38
rs771273943 Chr.6: 70234890-70234890 on Build GRCh38
rs886061696 Chr.6: 70234914-70234914 on Build GRCh38
rs186821588 Chr.6: 70234826-70234826 on Build GRCh38
rs201479028 Chr.6: 70234812-70234812 on Build GRCh38
rs776778931 Chr.6: 70234747-70234747 on Build GRCh38
rs766190162 Chr.6: 70234949-70234949 on Build GRCh38
rs148695653 Chr.6: 70234850-70234850 on Build GRCh38
rs200418420 Chr.6: 70234753-70234753 on Build GRCh38
rs1029245584 Chr.6: 70234870-70234870 on Build GRCh38
rs1005817817 Chr.6: 70234848-70234848 on Build GRCh38
rs955449925 Chr.6: 70234903-70234903 on Build GRCh38
rs763616322 Chr.6: 70234795-70234795 on Build GRCh38
rs373480203 Chr.6: 70234787-70234787 on Build GRCh38
rs1032780767 Chr.6: 70234921-70234921 on Build GRCh38
rs10945205 Chr.6: 70234739-70234739 on Build GRCh38
rs761379477 Chr.6: 70234922-70234922 on Build GRCh38
rs774807480 Chr.6: 70234891-70234891 on Build GRCh38
rs751249539 Chr.6: 70234821-70234821 on Build GRCh38
rs771313083 Chr.6: 70234786-70234786 on Build GRCh38
rs977443998 Chr.6: 70234907-70234907 on Build GRCh38
rs997685722 Chr.6: 70234854-70234854 on Build GRCh38
rs137958247 Chr.6: 70234804-70234804 on Build GRCh38
rs1049240844 Chr.6: 70234799-70234799 on Build GRCh38
rs781246762 Chr.6: 70234825-70234825 on Build GRCh38
rs748275516 Chr.6: 70234847-70234847 on Build GRCh38
rs894732182 Chr.6: 70234754-70234754 on Build GRCh38
rs765737002 Chr.6: 70234830-70234831 on Build GRCh38
rs143853613 Chr.6: 70234817-70234817 on Build GRCh38
rs769986009 Chr.6: 70234849-70234849 on Build GRCh38
rs760378269 Chr.6: 70234794-70234794 on Build GRCh38
rs867499190 Chr.6: 70234838-70234838 on Build GRCh38
rs953644315 Chr.6: 70234883-70234883 on Build GRCh38
rs750991236 Chr.6: 70234895-70234895 on Build GRCh38
rs769565735 Chr.6: 70234735-70234735 on Build GRCh38
rs988603716 Chr.6: 70234957-70234957 on Build GRCh38
rs753370035 Chr.6: 70234800-70234800 on Build GRCh38
rs936152967 Chr.6: 70234933-70234933 on Build GRCh38
rs776235308 Chr.6: 70234912-70234912 on Build GRCh38
rs12191701 Chr.6: 70234762-70234762 on Build GRCh38

View Full Product Details