Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 47362289-47362550 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHX34
Entrez Gene ID
9704
Gene Name
DEAH-box helicase 34
Gene Aliases
DDX34, HRH1
UniGene
Hs.151706
Gene Chromosome Location
Chr.19: 47349281-47382704 on Build GRCh38
Transcripts
NM_014681.5 XM_005259443.3 XM_011527550.2 XM_011527551.2 XM_017027524.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
998712 c.1420C>T p.R474W Chr.19: 47362520-47362520 on Build GRCh38
3106276 c.1414C>T p.L472L Chr.19: 47362514-47362514 on Build GRCh38
5981573 c.1399G>T p.D467Y Chr.19: 47362499-47362499 on Build GRCh38
6891521 c.1428G>A p.Q476Q Chr.19: 47362528-47362528 on Build GRCh38
3396349 c.1397A>G p.Y466C Chr.19: 47362497-47362497 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs772893244 Chr.19: 47362523-47362523 on Build GRCh38
rs568188204 Chr.19: 47362355-47362355 on Build GRCh38
rs200925941 Chr.19: 47362504-47362504 on Build GRCh38
rs955666138 Chr.19: 47362470-47362470 on Build GRCh38
rs371716085 Chr.19: 47362445-47362446 on Build GRCh38
rs867020512 Chr.19: 47362411-47362411 on Build GRCh38
rs750241552 Chr.19: 47362442-47362442 on Build GRCh38
rs370413988 Chr.19: 47362457-47362457 on Build GRCh38
rs762759611 Chr.19: 47362445-47362445 on Build GRCh38
rs754050697 Chr.19: 47362461-47362461 on Build GRCh38
rs550355621 Chr.19: 47362396-47362396 on Build GRCh38
rs1051022169 Chr.19: 47362358-47362358 on Build GRCh38
rs151026907 Chr.19: 47362413-47362413 on Build GRCh38
rs906355252 Chr.19: 47362436-47362436 on Build GRCh38
rs377348757 Chr.19: 47362495-47362495 on Build GRCh38
rs572752500 Chr.19: 47362417-47362417 on Build GRCh38
rs373031819 Chr.19: 47362440-47362440 on Build GRCh38
rs368316466 Chr.19: 47362490-47362490 on Build GRCh38
rs756208168 Chr.19: 47362458-47362458 on Build GRCh38
rs377016600 Chr.19: 47362455-47362455 on Build GRCh38
rs140363128 Chr.19: 47362498-47362498 on Build GRCh38
rs371265202 Chr.19: 47362429-47362429 on Build GRCh38
rs536746285 Chr.19: 47362394-47362394 on Build GRCh38
rs374868635 Chr.19: 47362463-47362463 on Build GRCh38
rs369444024 Chr.19: 47362503-47362503 on Build GRCh38
rs576592618 Chr.19: 47362514-47362514 on Build GRCh38
rs1040406505 Chr.19: 47362410-47362410 on Build GRCh38
rs747897472 Chr.19: 47362381-47362381 on Build GRCh38
rs769467907 Chr.19: 47362520-47362520 on Build GRCh38
rs535049999 Chr.19: 47362430-47362430 on Build GRCh38
rs547861516 Chr.19: 47362342-47362342 on Build GRCh38
rs746207981 Chr.19: 47362491-47362491 on Build GRCh38
rs778067783 Chr.19: 47362460-47362460 on Build GRCh38
rs539358785 Chr.19: 47362414-47362414 on Build GRCh38
rs1023218878 Chr.19: 47362448-47362448 on Build GRCh38
rs540826341 Chr.19: 47362421-47362452 on Build GRCh38
rs761458651 Chr.19: 47362431-47362431 on Build GRCh38
rs138130970 Chr.19: 47362509-47362509 on Build GRCh38
rs191810019 Chr.19: 47362415-47362415 on Build GRCh38
rs759079177 Chr.19: 47362439-47362440 on Build GRCh38
rs764952875 Chr.19: 47362435-47362435 on Build GRCh38
rs145555932 Chr.19: 47362499-47362499 on Build GRCh38

View Full Product Details