Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 218661560-218661809 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCS1L
Entrez Gene ID
617
Gene Name
BCS1 homolog, ubiquinol-cytochrome c reductase complex chaperone
Gene Aliases
BCS, BCS1, BJS, FLNMS, GRACILE, Hs.6719, MC3DN1, PTD, h-BCS, h-BCS1
UniGene
Hs.471401
Gene Chromosome Location
Chr.2: 218659656-218663443 on Build GRCh38
Transcripts
NM_001079866.1 NM_001257342.1 NM_001257343.1 NM_001257344.1 NM_001318836.1 NM_001320717.1 NM_004328.4 XM_005246748.2 XM_005246749.3 XM_006712678.1 XM_017004631.1 XM_017004632.1 XM_017004633.1 XM_017004634.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4518024 c.497_498CC>TT p.T166I Chr.2: 218661795-218661796 on Build GRCh38
309464 c.463C>G p.R155G Chr.2: 218661761-218661761 on Build GRCh38
6031008 c.483G>A p.Q161Q Chr.2: 218661781-218661781 on Build GRCh38
4185643 c.464_465delGA p.E156fs*19 Chr.2: 218661762-218661763 on Build GRCh38
6611135 c.498C>T p.T166T Chr.2: 218661796-218661796 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs987238654 Chr.2: 218661759-218661759 on Build GRCh38
rs1001355643 Chr.2: 218661677-218661677 on Build GRCh38
rs982826850 Chr.2: 218661603-218661603 on Build GRCh38
rs762566470 Chr.2: 218661641-218661641 on Build GRCh38
rs542467112 Chr.2: 218661636-218661636 on Build GRCh38
rs1032863083 Chr.2: 218661697-218661697 on Build GRCh38
rs769144065 Chr.2: 218661752-218661752 on Build GRCh38
rs767028489 Chr.2: 218661744-218661746 on Build GRCh38
rs751468984 Chr.2: 218661724-218661724 on Build GRCh38
rs1047358116 Chr.2: 218661612-218661612 on Build GRCh38
rs905811405 Chr.2: 218661660-218661661 on Build GRCh38
rs774688562 Chr.2: 218661761-218661761 on Build GRCh38
rs758462006 Chr.2: 218661591-218661591 on Build GRCh38
rs543586069 Chr.2: 218661703-218661703 on Build GRCh38
rs757128513 Chr.2: 218661647-218661647 on Build GRCh38
rs777682063 Chr.2: 218661717-218661717 on Build GRCh38
rs770698586 Chr.2: 218661748-218661750 on Build GRCh38
rs915709226 Chr.2: 218661736-218661736 on Build GRCh38
rs745537383 Chr.2: 218661751-218661751 on Build GRCh38
rs373105002 Chr.2: 218661776-218661776 on Build GRCh38
rs879252201 Chr.2: 218661624-218661624 on Build GRCh38
rs879132075 Chr.2: 218661646-218661646 on Build GRCh38
rs752635954 Chr.2: 218661579-218661579 on Build GRCh38
rs779638704 Chr.2: 218661754-218661754 on Build GRCh38
rs368868835 Chr.2: 218661743-218661743 on Build GRCh38
rs531442370 Chr.2: 218661656-218661656 on Build GRCh38
rs1042987872 Chr.2: 218661785-218661785 on Build GRCh38
rs121908573 Chr.2: 218661762-218661762 on Build GRCh38
rs769187920 Chr.2: 218661755-218661755 on Build GRCh38
rs746903284 Chr.2: 218661738-218661738 on Build GRCh38
rs930414096 Chr.2: 218661611-218661611 on Build GRCh38
rs777357804 Chr.2: 218661741-218661743 on Build GRCh38
rs532199775 Chr.2: 218661744-218661744 on Build GRCh38
rs780651750 Chr.2: 218661750-218661750 on Build GRCh38
rs560845438 Chr.2: 218661640-218661640 on Build GRCh38
rs780002216 Chr.2: 218661733-218661733 on Build GRCh38

View Full Product Details