Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 24064182-24064407 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2S3
Entrez Gene ID
1968
Gene Name
eukaryotic translation initiation factor 2 subunit gamma
Gene Aliases
EIF2, EIF2G, EIF2gamma, eIF-2gA
UniGene
Hs.539684
Gene Chromosome Location
Chr.X: 24054948-24078810 on Build GRCh38
Transcripts
NM_001415.3

SNP Detail

SNP ID Chromosome Location
rs886040855 Chr.X: 24064228-24064228 on Build GRCh38
rs772198732 Chr.X: 24064359-24064359 on Build GRCh38
rs188182089 Chr.X: 24064360-24064360 on Build GRCh38
rs777262915 Chr.X: 24064359-24064360 on Build GRCh38
rs768460610 Chr.X: 24064379-24064379 on Build GRCh38
rs760397776 Chr.X: 24064364-24064364 on Build GRCh38
rs778615116 Chr.X: 24064238-24064238 on Build GRCh38
rs748928414 Chr.X: 24064224-24064224 on Build GRCh38
rs909247776 Chr.X: 24064369-24064369 on Build GRCh38
rs372407051 Chr.X: 24064310-24064310 on Build GRCh38
rs770585940 Chr.X: 24064226-24064226 on Build GRCh38
rs919808510 Chr.X: 24064303-24064303 on Build GRCh38

View Full Product Details