Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 111516059-111516328 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf88
Entrez Gene ID
399949
Gene Name
chromosome 11 open reading frame 88
Gene Aliases
-
UniGene
Hs.411600
Gene Chromosome Location
Chr.11: 111514785-111537031 on Build GRCh38
Transcripts
NM_001100388.1 NM_207430.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6025240 c.288C>T p.A96A Chr.11: 111516059-111516059 on Build GRCh38
259708 c.333C>T p.L111L Chr.11: 111516104-111516104 on Build GRCh38
1289470 c.295C>A p.L99I Chr.11: 111516066-111516066 on Build GRCh38
7115947 c.313G>C p.E105Q Chr.11: 111516084-111516084 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs745832061 Chr.11: 111516132-111516132 on Build GRCh38
rs548904308 Chr.11: 111516240-111516240 on Build GRCh38
rs138544112 Chr.11: 111516284-111516284 on Build GRCh38
rs896335456 Chr.11: 111516202-111516202 on Build GRCh38
rs559925272 Chr.11: 111516178-111516178 on Build GRCh38
rs928049665 Chr.11: 111516114-111516114 on Build GRCh38
rs758180569 Chr.11: 111516134-111516134 on Build GRCh38
rs527437968 Chr.11: 111516181-111516181 on Build GRCh38
rs778936446 Chr.11: 111516102-111516102 on Build GRCh38
rs1056732123 Chr.11: 111516187-111516187 on Build GRCh38
rs960663846 Chr.11: 111516162-111516162 on Build GRCh38
rs761277662 Chr.11: 111516126-111516128 on Build GRCh38
rs187949187 Chr.11: 111516272-111516272 on Build GRCh38
rs773563983 Chr.11: 111516135-111516135 on Build GRCh38
rs905981731 Chr.11: 111516177-111516177 on Build GRCh38
rs182486763 Chr.11: 111516098-111516098 on Build GRCh38
rs531293379 Chr.11: 111516254-111516254 on Build GRCh38
rs900937552 Chr.11: 111516296-111516296 on Build GRCh38
rs766568527 Chr.11: 111516180-111516180 on Build GRCh38
rs913150696 Chr.11: 111516198-111516198 on Build GRCh38
rs978989661 Chr.11: 111516245-111516245 on Build GRCh38
rs370039147 Chr.11: 111516253-111516253 on Build GRCh38
rs567397545 Chr.11: 111516241-111516241 on Build GRCh38
rs763649488 Chr.11: 111516085-111516085 on Build GRCh38
rs549779277 Chr.11: 111516256-111516256 on Build GRCh38
rs946019689 Chr.11: 111516208-111516208 on Build GRCh38

View Full Product Details