Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 9993094-9993341 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC12A
Entrez Gene ID
160364
Gene Name
C-type lectin domain family 12 member A
Gene Aliases
CD371, CLL-1, CLL1, DCAL-2, MICL
UniGene
Hs.190519
Gene Chromosome Location
Chr.12: 9951316-9995209 on Build GRCh38
Transcripts
XM_006719035.3 XM_011520573.2 NM_001099431.1 NM_016509.3 XM_005253382.4 XM_011520685.2 XM_011520686.2 XM_017019395.1 XM_017019396.1

Gene Symbol
CLEC1B
Entrez Gene ID
51266
Gene Name
C-type lectin domain family 1 member B
Gene Aliases
1810061I13Rik, CLEC2, CLEC2B, PRO1384, QDED721
UniGene
Hs.409794
Gene Chromosome Location
Chr.12: 9986119-10001894 on Build GRCh38
Transcripts
XM_006719035.3 XM_011520573.2 NM_001099431.1 NM_016509.3 XM_005253382.4 XM_011520685.2 XM_011520686.2 XM_017019395.1 XM_017019396.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7133483 c.651G>C p.E217D Chr.12: 9993182-9993182 on Build GRCh38
1746668 c.290-1G>A p.? Chr.12: 9993271-9993271 on Build GRCh38
7312607 c.340C>A p.P114T Chr.12: 9993220-9993220 on Build GRCh38
4553852 c.603G>A p.G201G Chr.12: 9993230-9993230 on Build GRCh38
260133 c.301A>C p.N101H Chr.12: 9993259-9993259 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777285402 Chr.12: 9993292-9993292 on Build GRCh38
rs1053420338 Chr.12: 9993167-9993167 on Build GRCh38
rs375345439 Chr.12: 9993150-9993150 on Build GRCh38
rs756654849 Chr.12: 9993275-9993275 on Build GRCh38
rs868490047 Chr.12: 9993248-9993248 on Build GRCh38
rs1035753548 Chr.12: 9993238-9993238 on Build GRCh38
rs760320045 Chr.12: 9993189-9993189 on Build GRCh38
rs894518759 Chr.12: 9993242-9993242 on Build GRCh38
rs765374313 Chr.12: 9993244-9993244 on Build GRCh38
rs771574163 Chr.12: 9993179-9993179 on Build GRCh38
rs781616867 Chr.12: 9993154-9993154 on Build GRCh38
rs781594653 Chr.12: 9993269-9993269 on Build GRCh38
rs772928618 Chr.12: 9993188-9993188 on Build GRCh38
rs1041909532 Chr.12: 9993219-9993219 on Build GRCh38
rs757614783 Chr.12: 9993260-9993260 on Build GRCh38
rs757786077 Chr.12: 9993271-9993271 on Build GRCh38
rs770915236 Chr.12: 9993194-9993194 on Build GRCh38
rs746441331 Chr.12: 9993161-9993161 on Build GRCh38
rs867154854 Chr.12: 9993262-9993262 on Build GRCh38
rs202115320 Chr.12: 9993168-9993168 on Build GRCh38
rs15635 Chr.12: 9993132-9993132 on Build GRCh38
rs752989413 Chr.12: 9993145-9993145 on Build GRCh38
rs371450388 Chr.12: 9993287-9993287 on Build GRCh38
rs180901380 Chr.12: 9993236-9993236 on Build GRCh38
rs758584854 Chr.12: 9993134-9993134 on Build GRCh38
rs771615733 Chr.12: 9993288-9993288 on Build GRCh38
rs776692567 Chr.12: 9993222-9993222 on Build GRCh38
rs752683376 Chr.12: 9993245-9993245 on Build GRCh38
rs199859311 Chr.12: 9993162-9993162 on Build GRCh38
rs756656317 Chr.12: 9993265-9993266 on Build GRCh38
rs535930013 Chr.12: 9993257-9993257 on Build GRCh38
rs746730744 Chr.12: 9993298-9993298 on Build GRCh38
rs750909097 Chr.12: 9993272-9993272 on Build GRCh38
rs188574713 Chr.12: 9993140-9993140 on Build GRCh38
rs764169274 Chr.12: 9993256-9993256 on Build GRCh38
rs763118730 Chr.12: 9993246-9993246 on Build GRCh38
rs368407925 Chr.12: 9993126-9993126 on Build GRCh38
rs567272126 Chr.12: 9993170-9993170 on Build GRCh38
rs1040914016 Chr.12: 9993131-9993131 on Build GRCh38
rs747805854 Chr.12: 9993172-9993172 on Build GRCh38
rs903346485 Chr.12: 9993220-9993220 on Build GRCh38
rs751824512 Chr.12: 9993148-9993148 on Build GRCh38
rs780560552 Chr.12: 9993276-9993276 on Build GRCh38

View Full Product Details