Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 117382837-117383068 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASZ1
Entrez Gene ID
136991
Gene Name
ankyrin repeat, SAM and basic leucine zipper domain containing 1
Gene Aliases
ALP1, ANKL1, C7orf7, CT1.19, GASZ, Orf3
UniGene
Hs.352412
Gene Chromosome Location
Chr.7: 117363222-117427523 on Build GRCh38
Transcripts
NM_001301821.1 NM_001301822.1 NM_130768.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6217226 c.745A>C p.T249P Chr.7: 117383053-117383053 on Build GRCh38
6606343 c.785C>G p.S262C Chr.7: 117383013-117383013 on Build GRCh38
1699239 c.802C>T p.H268Y Chr.7: 117382996-117382996 on Build GRCh38
7159381 c.772T>C p.L258L Chr.7: 117383026-117383026 on Build GRCh38
7356751 c.812G>T p.S271I Chr.7: 117382986-117382986 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs116289816 Chr.7: 117382928-117382928 on Build GRCh38
rs761978274 Chr.7: 117382977-117382977 on Build GRCh38
rs568708136 Chr.7: 117382986-117382986 on Build GRCh38
rs552398545 Chr.7: 117382900-117382900 on Build GRCh38
rs953078247 Chr.7: 117382880-117382880 on Build GRCh38
rs924173550 Chr.7: 117382936-117382936 on Build GRCh38
rs756486342 Chr.7: 117382985-117382985 on Build GRCh38
rs145217294 Chr.7: 117382901-117382901 on Build GRCh38
rs775345717 Chr.7: 117382964-117382964 on Build GRCh38
rs778933484 Chr.7: 117382921-117382921 on Build GRCh38
rs767760188 Chr.7: 117382979-117382979 on Build GRCh38
rs992255906 Chr.7: 117382963-117382963 on Build GRCh38
rs373634803 Chr.7: 117382980-117382980 on Build GRCh38
rs751655914 Chr.7: 117382971-117382971 on Build GRCh38
rs899021599 Chr.7: 117382994-117382994 on Build GRCh38
rs768791178 Chr.7: 117382927-117382927 on Build GRCh38
rs769745454 Chr.7: 117382953-117382953 on Build GRCh38
rs746147401 Chr.7: 117382969-117382970 on Build GRCh38
rs745783535 Chr.7: 117382925-117382925 on Build GRCh38
rs1023474540 Chr.7: 117383037-117383037 on Build GRCh38
rs887677206 Chr.7: 117382995-117382995 on Build GRCh38
rs1034788519 Chr.7: 117382894-117382894 on Build GRCh38
rs1036144827 Chr.7: 117383015-117383015 on Build GRCh38
rs756234130 Chr.7: 117383000-117383000 on Build GRCh38
rs754370827 Chr.7: 117382997-117382997 on Build GRCh38
rs368973451 Chr.7: 117382950-117382950 on Build GRCh38
rs76241408 Chr.7: 117382965-117382965 on Build GRCh38
rs764089881 Chr.7: 117382970-117382970 on Build GRCh38
rs373380517 Chr.7: 117382954-117382954 on Build GRCh38
rs779960662 Chr.7: 117383002-117383007 on Build GRCh38

View Full Product Details