Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 20133844-20134079 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF1AX
Entrez Gene ID
1964
Gene Name
eukaryotic translation initiation factor 1A, X-linked
Gene Aliases
EIF1A, EIF1AP1, EIF4C, eIF-1A, eIF-4C
UniGene
Hs.522590
Gene Chromosome Location
Chr.X: 20124518-20141848 on Build GRCh38
Transcripts
NM_001412.3

SNP Detail

SNP ID Chromosome Location
rs778931762 Chr.X: 20134029-20134029 on Build GRCh38
rs745610158 Chr.X: 20134030-20134030 on Build GRCh38
rs752396421 Chr.X: 20133923-20133923 on Build GRCh38
rs759546126 Chr.X: 20134014-20134014 on Build GRCh38
rs376252334 Chr.X: 20133917-20133917 on Build GRCh38
rs187186325 Chr.X: 20134031-20134031 on Build GRCh38
rs772100699 Chr.X: 20133941-20133941 on Build GRCh38
rs763662392 Chr.X: 20133968-20133968 on Build GRCh38
rs1049451175 Chr.X: 20134048-20134048 on Build GRCh38
rs574396802 Chr.X: 20133933-20133933 on Build GRCh38
rs755609734 Chr.X: 20133940-20133940 on Build GRCh38
rs932422100 Chr.X: 20133934-20133934 on Build GRCh38
rs781538389 Chr.X: 20134047-20134047 on Build GRCh38
rs774534184 Chr.X: 20133916-20133916 on Build GRCh38
rs767046516 Chr.X: 20133920-20133920 on Build GRCh38

View Full Product Details