Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 39734096-39734367 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLC
Entrez Gene ID
1178
Gene Name
Charcot-Leyden crystal galectin
Gene Aliases
GAL10, Gal-10, LGALS10, LGALS10A, LPPL_HUMAN
UniGene
Hs.889
Gene Chromosome Location
Chr.19: 39731253-39738029 on Build GRCh38
Transcripts
NM_001828.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5942816 c.253G>A p.D85N Chr.19: 39734333-39734333 on Build GRCh38
996415 c.237G>T p.K79N Chr.19: 39734349-39734349 on Build GRCh38
3533835 c.268G>A p.E90K Chr.19: 39734318-39734318 on Build GRCh38
1129862 c.240T>C p.N80N Chr.19: 39734346-39734346 on Build GRCh38
4819398 c.250C>T p.Q84* Chr.19: 39734336-39734336 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867849565 Chr.19: 39734336-39734336 on Build GRCh38
rs755688084 Chr.19: 39734239-39734239 on Build GRCh38
rs391646 Chr.19: 39734229-39734229 on Build GRCh38
rs760849445 Chr.19: 39734263-39734263 on Build GRCh38
rs777509854 Chr.19: 39734240-39734240 on Build GRCh38
rs375539895 Chr.19: 39734298-39734298 on Build GRCh38
rs776936554 Chr.19: 39734281-39734281 on Build GRCh38
rs561345244 Chr.19: 39734131-39734131 on Build GRCh38
rs984347826 Chr.19: 39734188-39734188 on Build GRCh38
rs1056626530 Chr.19: 39734228-39734228 on Build GRCh38
rs750873907 Chr.19: 39734293-39734293 on Build GRCh38
rs73559726 Chr.19: 39734312-39734312 on Build GRCh38
rs391333 Chr.19: 39734157-39734157 on Build GRCh38
rs753530424 Chr.19: 39734328-39734328 on Build GRCh38
rs529249496 Chr.19: 39734332-39734332 on Build GRCh38
rs985276210 Chr.19: 39734324-39734324 on Build GRCh38
rs953862011 Chr.19: 39734318-39734318 on Build GRCh38
rs948224532 Chr.19: 39734246-39734246 on Build GRCh38
rs756972983 Chr.19: 39734329-39734329 on Build GRCh38
rs770723553 Chr.19: 39734248-39734248 on Build GRCh38
rs779666817 Chr.19: 39734185-39734185 on Build GRCh38
rs762028295 Chr.19: 39734283-39734283 on Build GRCh38
rs768721064 Chr.19: 39734266-39734266 on Build GRCh38
rs767032975 Chr.19: 39734301-39734301 on Build GRCh38
rs1019617155 Chr.19: 39734136-39734136 on Build GRCh38
rs200169462 Chr.19: 39734322-39734322 on Build GRCh38
rs773634243 Chr.19: 39734285-39734285 on Build GRCh38
rs1042423682 Chr.19: 39734187-39734187 on Build GRCh38
rs391660 Chr.19: 39734247-39734247 on Build GRCh38
rs752104928 Chr.19: 39734302-39734302 on Build GRCh38
rs748092137 Chr.19: 39734266-39734266 on Build GRCh38
rs566785841 Chr.19: 39734260-39734260 on Build GRCh38
rs453827 Chr.19: 39734261-39734261 on Build GRCh38
rs965463244 Chr.19: 39734141-39734141 on Build GRCh38
rs384138 Chr.19: 39734346-39734346 on Build GRCh38
rs572052665 Chr.19: 39734163-39734163 on Build GRCh38
rs143109041 Chr.19: 39734345-39734345 on Build GRCh38
rs2074927 Chr.19: 39734254-39734254 on Build GRCh38
rs937780151 Chr.19: 39734303-39734303 on Build GRCh38
rs921117100 Chr.19: 39734170-39734170 on Build GRCh38
rs375867 Chr.19: 39734193-39734193 on Build GRCh38
rs528996376 Chr.19: 39734191-39734191 on Build GRCh38
rs939626911 Chr.19: 39734201-39734201 on Build GRCh38
rs375580 Chr.19: 39734159-39734159 on Build GRCh38
rs373288158 Chr.19: 39734255-39734255 on Build GRCh38

View Full Product Details