Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 37590230-37590473 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C4orf19
Entrez Gene ID
55286
Gene Name
chromosome 4 open reading frame 19
Gene Aliases
-
UniGene
Hs.107527
Gene Chromosome Location
Chr.4: 37453454-37596146 on Build GRCh38
Transcripts
NM_001104629.1 NM_018302.2 XM_011513712.2 XM_011513713.2 XM_017008361.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6026966 c.257A>C p.Q86P Chr.4: 37590312-37590312 on Build GRCh38
7240990 c.345C>G p.P115P Chr.4: 37590400-37590400 on Build GRCh38
222908 c.370G>A p.A124T Chr.4: 37590425-37590425 on Build GRCh38
1618785 c.245G>A p.G82E Chr.4: 37590300-37590300 on Build GRCh38
4124489 c.392C>T p.P131L Chr.4: 37590447-37590447 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766161298 Chr.4: 37590320-37590320 on Build GRCh38
rs749021414 Chr.4: 37590353-37590353 on Build GRCh38
rs140208747 Chr.4: 37590352-37590352 on Build GRCh38
rs143231920 Chr.4: 37590333-37590333 on Build GRCh38
rs968004040 Chr.4: 37590310-37590310 on Build GRCh38
rs780803730 Chr.4: 37590413-37590413 on Build GRCh38
rs747119173 Chr.4: 37590345-37590345 on Build GRCh38
rs747274913 Chr.4: 37590264-37590264 on Build GRCh38
rs749232596 Chr.4: 37590416-37590417 on Build GRCh38
rs763876800 Chr.4: 37590285-37590285 on Build GRCh38
rs761858334 Chr.4: 37590358-37590358 on Build GRCh38
rs371058859 Chr.4: 37590420-37590420 on Build GRCh38
rs774403294 Chr.4: 37590289-37590289 on Build GRCh38
rs766073458 Chr.4: 37590397-37590397 on Build GRCh38
rs754189646 Chr.4: 37590321-37590321 on Build GRCh38
rs914049776 Chr.4: 37590451-37590451 on Build GRCh38
rs139532209 Chr.4: 37590304-37590304 on Build GRCh38
rs752488878 Chr.4: 37590278-37590279 on Build GRCh38
rs35691133 Chr.4: 37590280-37590281 on Build GRCh38
rs975861402 Chr.4: 37590348-37590348 on Build GRCh38
rs779277111 Chr.4: 37590334-37590334 on Build GRCh38
rs544212016 Chr.4: 37590386-37590386 on Build GRCh38
rs145167191 Chr.4: 37590356-37590356 on Build GRCh38
rs753317803 Chr.4: 37590338-37590338 on Build GRCh38
rs762739859 Chr.4: 37590281-37590281 on Build GRCh38
rs775221705 Chr.4: 37590280-37590280 on Build GRCh38
rs201239178 Chr.4: 37590362-37590362 on Build GRCh38
rs760268266 Chr.4: 37590388-37590388 on Build GRCh38
rs753126994 Chr.4: 37590448-37590448 on Build GRCh38
rs758380758 Chr.4: 37590343-37590343 on Build GRCh38
rs756131822 Chr.4: 37590329-37590330 on Build GRCh38
rs35633618 Chr.4: 37590344-37590344 on Build GRCh38
rs62299700 Chr.4: 37590422-37590422 on Build GRCh38
rs114589329 Chr.4: 37590453-37590453 on Build GRCh38
rs984487767 Chr.4: 37590359-37590359 on Build GRCh38
rs771160913 Chr.4: 37590275-37590275 on Build GRCh38
rs750078312 Chr.4: 37590319-37590319 on Build GRCh38
rs772157970 Chr.4: 37590372-37590372 on Build GRCh38
rs765677047 Chr.4: 37590446-37590446 on Build GRCh38
rs767094811 Chr.4: 37590313-37590313 on Build GRCh38

View Full Product Details