Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 150795398-150795671 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMIM3
Entrez Gene ID
85027
Gene Name
small integral membrane protein 3
Gene Aliases
C5orf62, MST150, NID67
UniGene
Hs.29444
Gene Chromosome Location
Chr.5: 150777946-150796736 on Build GRCh38
Transcripts
NM_032947.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3702754 c.42C>A p.P14P Chr.5: 150795482-150795482 on Build GRCh38
6546811 c.27G>T p.M9I Chr.5: 150795467-150795467 on Build GRCh38
5890070 c.28G>A p.E10K Chr.5: 150795468-150795468 on Build GRCh38
5402729 c.12C>T p.V4V Chr.5: 150795452-150795452 on Build GRCh38
3827431 c.88A>C p.T30P Chr.5: 150795528-150795528 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs751458789 Chr.5: 150795630-150795630 on Build GRCh38
rs182871851 Chr.5: 150795589-150795589 on Build GRCh38
rs773230066 Chr.5: 150795610-150795610 on Build GRCh38
rs537962884 Chr.5: 150795535-150795535 on Build GRCh38
rs750420133 Chr.5: 150795554-150795554 on Build GRCh38
rs139420502 Chr.5: 150795586-150795586 on Build GRCh38
rs752775488 Chr.5: 150795550-150795550 on Build GRCh38
rs767506186 Chr.5: 150795487-150795487 on Build GRCh38
rs766544899 Chr.5: 150795504-150795504 on Build GRCh38
rs777726493 Chr.5: 150795436-150795436 on Build GRCh38
rs61736140 Chr.5: 150795547-150795547 on Build GRCh38
rs745884586 Chr.5: 150795592-150795592 on Build GRCh38
rs760628507 Chr.5: 150795601-150795601 on Build GRCh38
rs747661821 Chr.5: 150795584-150795584 on Build GRCh38
rs766276561 Chr.5: 150795625-150795625 on Build GRCh38
rs749203291 Chr.5: 150795437-150795437 on Build GRCh38
rs751811120 Chr.5: 150795484-150795484 on Build GRCh38
rs890825598 Chr.5: 150795518-150795518 on Build GRCh38
rs778753787 Chr.5: 150795499-150795499 on Build GRCh38
rs777149434 Chr.5: 150795465-150795465 on Build GRCh38
rs768158128 Chr.5: 150795531-150795531 on Build GRCh38
rs751696937 Chr.5: 150795429-150795429 on Build GRCh38
rs766443207 Chr.5: 150795428-150795428 on Build GRCh38
rs754655098 Chr.5: 150795580-150795580 on Build GRCh38
rs756013041 Chr.5: 150795431-150795431 on Build GRCh38
rs771216252 Chr.5: 150795588-150795588 on Build GRCh38
rs746561064 Chr.5: 150795451-150795451 on Build GRCh38
rs757205523 Chr.5: 150795496-150795496 on Build GRCh38
rs751193101 Chr.5: 150795579-150795579 on Build GRCh38
rs535808685 Chr.5: 150795552-150795552 on Build GRCh38
rs746685675 Chr.5: 150795519-150795519 on Build GRCh38
rs369432252 Chr.5: 150795442-150795442 on Build GRCh38
rs780010907 Chr.5: 150795571-150795571 on Build GRCh38
rs570998197 Chr.5: 150795474-150795474 on Build GRCh38
rs546977535 Chr.5: 150795585-150795585 on Build GRCh38
rs779816744 Chr.5: 150795517-150795517 on Build GRCh38
rs779867339 Chr.5: 150795460-150795478 on Build GRCh38
rs753740962 Chr.5: 150795491-150795491 on Build GRCh38
rs757018297 Chr.5: 150795440-150795440 on Build GRCh38
rs780742167 Chr.5: 150795583-150795583 on Build GRCh38
rs142179677 Chr.5: 150795441-150795441 on Build GRCh38
rs759564022 Chr.5: 150795486-150795486 on Build GRCh38
rs768317242 Chr.5: 150795456-150795456 on Build GRCh38
rs922181099 Chr.5: 150795502-150795502 on Build GRCh38
rs888713023 Chr.5: 150795597-150795597 on Build GRCh38
rs764260778 Chr.5: 150795606-150795606 on Build GRCh38
rs762991132 Chr.5: 150795624-150795624 on Build GRCh38
rs574763074 Chr.5: 150795548-150795548 on Build GRCh38
rs924945076 Chr.5: 150795488-150795488 on Build GRCh38
rs188375591 Chr.5: 150795641-150795641 on Build GRCh38
rs554478573 Chr.5: 150795556-150795556 on Build GRCh38
rs773605312 Chr.5: 150795540-150795540 on Build GRCh38
rs749205561 Chr.5: 150795481-150795482 on Build GRCh38
rs2278396 Chr.5: 150795591-150795591 on Build GRCh38
rs758089499 Chr.5: 150795515-150795515 on Build GRCh38
rs763149754 Chr.5: 150795477-150795477 on Build GRCh38
rs766638068 Chr.5: 150795480-150795480 on Build GRCh38
rs543974134 Chr.5: 150795642-150795642 on Build GRCh38
rs534788836 Chr.5: 150795508-150795508 on Build GRCh38
rs372373149 Chr.5: 150795534-150795534 on Build GRCh38
rs780575873 Chr.5: 150795532-150795532 on Build GRCh38
rs779685832 Chr.5: 150795445-150795445 on Build GRCh38
rs754970465 Chr.5: 150795605-150795605 on Build GRCh38

View Full Product Details