Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 112163547-112163772 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HECTD4
Entrez Gene ID
283450
Gene Name
HECT domain E3 ubiquitin protein ligase 4
Gene Aliases
C12orf51, POTAGE
UniGene
Hs.530943
Gene Chromosome Location
Chr.12: 112160188-112382092 on Build GRCh38
Transcripts
NM_001109662.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4912888 c.11471G>A p.R3824Q Chr.12: 112163702-112163702 on Build GRCh38
1561835 c.11522G>A p.R3841Q Chr.12: 112163651-112163651 on Build GRCh38
3954189 c.11606C>T p.P3869L Chr.12: 112163567-112163567 on Build GRCh38
6202870 c.11559G>A p.L3853L Chr.12: 112163614-112163614 on Build GRCh38
1197985 c.11461G>A p.A3821T Chr.12: 112163712-112163712 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758133588 Chr.12: 112163715-112163715 on Build GRCh38
rs141828248 Chr.12: 112163629-112163629 on Build GRCh38
rs755661535 Chr.12: 112163655-112163655 on Build GRCh38
rs540965990 Chr.12: 112163719-112163719 on Build GRCh38
rs923427135 Chr.12: 112163672-112163672 on Build GRCh38
rs181634938 Chr.12: 112163651-112163651 on Build GRCh38
rs941681149 Chr.12: 112163703-112163703 on Build GRCh38
rs967065546 Chr.12: 112163721-112163721 on Build GRCh38
rs545772641 Chr.12: 112163687-112163687 on Build GRCh38
rs564542020 Chr.12: 112163702-112163702 on Build GRCh38
rs760968860 Chr.12: 112163641-112163641 on Build GRCh38
rs978323037 Chr.12: 112163750-112163750 on Build GRCh38
rs61748839 Chr.12: 112163713-112163713 on Build GRCh38
rs968039980 Chr.12: 112163634-112163634 on Build GRCh38
rs760008766 Chr.12: 112163716-112163716 on Build GRCh38
rs929500932 Chr.12: 112163677-112163677 on Build GRCh38
rs761302717 Chr.12: 112163669-112163669 on Build GRCh38
rs775441229 Chr.12: 112163588-112163588 on Build GRCh38
rs1018405682 Chr.12: 112163665-112163665 on Build GRCh38
rs905555813 Chr.12: 112163735-112163735 on Build GRCh38
rs535516940 Chr.12: 112163612-112163612 on Build GRCh38
rs573784336 Chr.12: 112163575-112163575 on Build GRCh38
rs377053677 Chr.12: 112163656-112163656 on Build GRCh38
rs910616854 Chr.12: 112163647-112163647 on Build GRCh38

View Full Product Details