Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 915985-916240 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANGPT4
Entrez Gene ID
51378
Gene Name
angiopoietin 4
Gene Aliases
ANG3, ANG4
UniGene
Hs.278973
Gene Chromosome Location
Chr.20: 869899-916367 on Build GRCh38
Transcripts
NM_001322809.1 NM_015985.3 XM_011529239.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
478419 c.225C>T p.N75N Chr.20: 915990-915990 on Build GRCh38
3841600 c.227C>A p.P76Q Chr.20: 915988-915988 on Build GRCh38
478420 c.148C>T p.P50S Chr.20: 916067-916067 on Build GRCh38
6599626 c.55A>G p.M19V Chr.20: 916160-916160 on Build GRCh38
2933889 c.82G>A p.E28K Chr.20: 916133-916133 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772883230 Chr.20: 916094-916094 on Build GRCh38
rs764585947 Chr.20: 916128-916128 on Build GRCh38
rs200815484 Chr.20: 916043-916043 on Build GRCh38
rs764806114 Chr.20: 916116-916116 on Build GRCh38
rs969626509 Chr.20: 916160-916160 on Build GRCh38
rs770645507 Chr.20: 916101-916101 on Build GRCh38
rs745899909 Chr.20: 916050-916050 on Build GRCh38
rs993180137 Chr.20: 916089-916089 on Build GRCh38
rs146594844 Chr.20: 916072-916072 on Build GRCh38
rs372508631 Chr.20: 916178-916178 on Build GRCh38
rs140900541 Chr.20: 916015-916015 on Build GRCh38
rs762278397 Chr.20: 916048-916048 on Build GRCh38
rs980098439 Chr.20: 916170-916170 on Build GRCh38
rs746957688 Chr.20: 916082-916082 on Build GRCh38
rs750615088 Chr.20: 916134-916134 on Build GRCh38
rs765788530 Chr.20: 916019-916019 on Build GRCh38
rs762045720 Chr.20: 916130-916130 on Build GRCh38
rs199886060 Chr.20: 916099-916099 on Build GRCh38
rs972144749 Chr.20: 916218-916219 on Build GRCh38
rs369053720 Chr.20: 916129-916129 on Build GRCh38
rs764341218 Chr.20: 916032-916032 on Build GRCh38
rs748015813 Chr.20: 916084-916084 on Build GRCh38
rs1020061490 Chr.20: 916026-916026 on Build GRCh38
rs771963050 Chr.20: 916087-916087 on Build GRCh38
rs774515703 Chr.20: 916042-916055 on Build GRCh38
rs17702306 Chr.20: 916110-916110 on Build GRCh38
rs752749565 Chr.20: 916162-916162 on Build GRCh38
rs758206666 Chr.20: 916184-916184 on Build GRCh38
rs200170732 Chr.20: 916147-916147 on Build GRCh38
rs776299477 Chr.20: 916106-916106 on Build GRCh38
rs776112183 Chr.20: 916118-916118 on Build GRCh38
rs767686759 Chr.20: 916131-916131 on Build GRCh38
rs144442482 Chr.20: 916045-916045 on Build GRCh38
rs746729787 Chr.20: 916196-916196 on Build GRCh38
rs953311251 Chr.20: 916074-916074 on Build GRCh38
rs753031251 Chr.20: 916025-916025 on Build GRCh38
rs777640954 Chr.20: 916191-916191 on Build GRCh38
rs201720367 Chr.20: 916029-916029 on Build GRCh38
rs756237515 Chr.20: 916145-916145 on Build GRCh38
rs776217811 Chr.20: 916010-916010 on Build GRCh38
rs757415116 Chr.20: 916044-916044 on Build GRCh38
rs778863210 Chr.20: 916083-916083 on Build GRCh38
rs763353908 Chr.20: 916122-916122 on Build GRCh38
rs746681989 Chr.20: 916095-916095 on Build GRCh38

View Full Product Details