Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 138813158-138813419 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ECT2L
Entrez Gene ID
345930
Gene Name
epithelial cell transforming 2 like
Gene Aliases
ARHGEF32, C6orf91, FBXO49, LFDH, dJ509I19.2, dJ509I19.3, dJ509I19.5
UniGene
Hs.660603
Gene Chromosome Location
Chr.6: 138796107-138904070 on Build GRCh38
Transcripts
NM_001077706.2 NM_001195037.2 XM_006715472.3 XM_011535795.2 XM_011535797.2 XM_017010828.1 XM_017010829.1 XM_017010830.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4909578 c.50A>T p.K17M Chr.6: 138813324-138813324 on Build GRCh38
1073858 c.8G>A p.S3N Chr.6: 138813282-138813282 on Build GRCh38
7111691 c.37C>G p.P13A Chr.6: 138813311-138813311 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs370601948 Chr.6: 138813241-138813241 on Build GRCh38
rs149487449 Chr.6: 138813378-138813378 on Build GRCh38
rs781779284 Chr.6: 138813334-138813334 on Build GRCh38
rs746231934 Chr.6: 138813392-138813392 on Build GRCh38
rs1024101412 Chr.6: 138813296-138813296 on Build GRCh38
rs373540636 Chr.6: 138813327-138813327 on Build GRCh38
rs778668888 Chr.6: 138813258-138813258 on Build GRCh38
rs753926308 Chr.6: 138813359-138813359 on Build GRCh38
rs371475573 Chr.6: 138813270-138813270 on Build GRCh38
rs764990525 Chr.6: 138813233-138813233 on Build GRCh38
rs771630511 Chr.6: 138813311-138813311 on Build GRCh38
rs765108112 Chr.6: 138813248-138813248 on Build GRCh38
rs768172622 Chr.6: 138813297-138813297 on Build GRCh38
rs377461352 Chr.6: 138813348-138813348 on Build GRCh38
rs771860037 Chr.6: 138813277-138813277 on Build GRCh38
rs369742195 Chr.6: 138813324-138813324 on Build GRCh38
rs758140303 Chr.6: 138813377-138813377 on Build GRCh38
rs764215943 Chr.6: 138813351-138813351 on Build GRCh38
rs763540724 Chr.6: 138813301-138813301 on Build GRCh38
rs776053139 Chr.6: 138813300-138813300 on Build GRCh38
rs779480754 Chr.6: 138813240-138813240 on Build GRCh38
rs79219465 Chr.6: 138813286-138813286 on Build GRCh38
rs757185495 Chr.6: 138813371-138813371 on Build GRCh38
rs753786785 Chr.6: 138813246-138813246 on Build GRCh38
rs1043300433 Chr.6: 138813381-138813381 on Build GRCh38
rs961044495 Chr.6: 138813315-138813315 on Build GRCh38
rs906935871 Chr.6: 138813362-138813362 on Build GRCh38
rs778756961 Chr.6: 138813375-138813375 on Build GRCh38

View Full Product Details