Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 155089188-155089439 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRCC3
Entrez Gene ID
79184
Gene Name
BRCA1/BRCA2-containing complex subunit 3
Gene Aliases
BRCC36, C6.1A, CXorf53
UniGene
Hs.558537
Gene Chromosome Location
Chr.X: 155071403-155126766 on Build GRCh38
Transcripts
NM_001018055.2 NM_001242640.1 NM_024332.3 XM_005274751.4 XM_017029838.1

SNP Detail

SNP ID Chromosome Location
rs782818838 Chr.X: 155089299-155089299 on Build GRCh38
rs1023064832 Chr.X: 155089374-155089374 on Build GRCh38
rs782639233 Chr.X: 155089238-155089238 on Build GRCh38
rs12833808 Chr.X: 155089288-155089288 on Build GRCh38
rs781946105 Chr.X: 155089395-155089395 on Build GRCh38
rs370260379 Chr.X: 155089384-155089384 on Build GRCh38
rs781952593 Chr.X: 155089367-155089368 on Build GRCh38
rs782237616 Chr.X: 155089231-155089231 on Build GRCh38
rs371170397 Chr.X: 155089225-155089225 on Build GRCh38
rs377415541 Chr.X: 155089273-155089273 on Build GRCh38
rs1012964975 Chr.X: 155089365-155089365 on Build GRCh38
rs957684045 Chr.X: 155089364-155089364 on Build GRCh38
rs782313824 Chr.X: 155089375-155089375 on Build GRCh38
rs782442985 Chr.X: 155089398-155089398 on Build GRCh38
rs371817972 Chr.X: 155089365-155089366 on Build GRCh38
rs781851163 Chr.X: 155089353-155089353 on Build GRCh38
rs782129717 Chr.X: 155089373-155089373 on Build GRCh38
rs201427873 Chr.X: 155089237-155089237 on Build GRCh38

View Full Product Details