Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 47129332-47129589 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC27
Entrez Gene ID
996
Gene Name
cell division cycle 27
Gene Aliases
ANAPC3, APC3, CDC27Hs, D0S1430E, D17S978E, H-NUC, HNUC, NUC2
UniGene
Hs.463295
Gene Chromosome Location
Chr.17: 47117697-47189332 on Build GRCh38
Transcripts
NM_001114091.2 NM_001256.4 NM_001293089.1 NM_001293091.1 XM_011525546.2 XM_011525547.2 XM_011525548.2 XM_011525549.2 XM_011525550.2 XM_017025481.1 XM_017025482.1 XM_017025483.1 XM_017025484.1 XM_017025485.1 XM_017025486.1 XM_017025487.1 XM_017025488.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6841787 c.2084C>A p.A695D Chr.17: 47129469-47129469 on Build GRCh38
6841786 c.2102C>A p.A701D Chr.17: 47129469-47129469 on Build GRCh38
980362 c.2121G>T p.K707N Chr.17: 47129450-47129450 on Build GRCh38
1710388 c.2072C>T p.S691L Chr.17: 47129499-47129499 on Build GRCh38
5010271 c.2155_2160+9delTATAAGGTAAGATAA p.? Chr.17: 47129384-47129398 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779450686 Chr.17: 47129538-47129538 on Build GRCh38
rs976146808 Chr.17: 47129551-47129551 on Build GRCh38
rs774750632 Chr.17: 47129372-47129372 on Build GRCh38
rs113085535 Chr.17: 47129417-47129417 on Build GRCh38
rs371098155 Chr.17: 47129380-47129380 on Build GRCh38
rs1006735569 Chr.17: 47129453-47129453 on Build GRCh38
rs766725312 Chr.17: 47129421-47129421 on Build GRCh38
rs773466519 Chr.17: 47129408-47129408 on Build GRCh38
rs772475829 Chr.17: 47129543-47129543 on Build GRCh38
rs1021579791 Chr.17: 47129396-47129396 on Build GRCh38
rs745395809 Chr.17: 47129361-47129361 on Build GRCh38
rs769332991 Chr.17: 47129535-47129535 on Build GRCh38
rs756844266 Chr.17: 47129510-47129510 on Build GRCh38
rs759723257 Chr.17: 47129437-47129437 on Build GRCh38
rs567090210 Chr.17: 47129557-47129557 on Build GRCh38
rs1016614216 Chr.17: 47129443-47129443 on Build GRCh38
rs753924753 Chr.17: 47129423-47129423 on Build GRCh38
rs753652128 Chr.17: 47129556-47129556 on Build GRCh38
rs752626597 Chr.17: 47129479-47129479 on Build GRCh38
rs751375797 Chr.17: 47129509-47129509 on Build GRCh38
rs532498880 Chr.17: 47129527-47129527 on Build GRCh38
rs761041557 Chr.17: 47129419-47129419 on Build GRCh38
rs777495484 Chr.17: 47129498-47129498 on Build GRCh38
rs112217389 Chr.17: 47129444-47129444 on Build GRCh38
rs781346820 Chr.17: 47129441-47129441 on Build GRCh38
rs201715412 Chr.17: 47129381-47129381 on Build GRCh38
rs761147763 Chr.17: 47129564-47129564 on Build GRCh38
rs200590868 Chr.17: 47129480-47129480 on Build GRCh38
rs368016453 Chr.17: 47129541-47129541 on Build GRCh38
rs780844929 Chr.17: 47129519-47129519 on Build GRCh38

View Full Product Details