Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 93242357-93242630 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTBD7
Entrez Gene ID
55727
Gene Name
BTB domain containing 7
Gene Aliases
FUP1
UniGene
Hs.525549
Gene Chromosome Location
Chr.14: 93237550-93333092 on Build GRCh38
Transcripts
NM_001002860.3 NM_001289133.1 XM_011536939.2 XM_017021438.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1371643 c.3194C>T p.T1065I Chr.14: 93242478-93242478 on Build GRCh38
6613405 c.2204G>A p.R735K Chr.14: 93242415-93242415 on Build GRCh38
3499057 c.3119C>T p.P1040L Chr.14: 93242553-93242553 on Build GRCh38
7416414 c.2222C>T p.A741V Chr.14: 93242397-93242397 on Build GRCh38
7273066 c.2086G>A p.A696T Chr.14: 93242533-93242533 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775432782 Chr.14: 93242393-93242393 on Build GRCh38
rs961114841 Chr.14: 93242498-93242498 on Build GRCh38
rs772822976 Chr.14: 93242508-93242509 on Build GRCh38
rs61740741 Chr.14: 93242554-93242554 on Build GRCh38
rs146841523 Chr.14: 93242535-93242535 on Build GRCh38
rs767026101 Chr.14: 93242511-93242511 on Build GRCh38
rs202055956 Chr.14: 93242464-93242464 on Build GRCh38
rs768644384 Chr.14: 93242505-93242505 on Build GRCh38
rs747016085 Chr.14: 93242487-93242487 on Build GRCh38
rs368457475 Chr.14: 93242601-93242601 on Build GRCh38
rs754402000 Chr.14: 93242444-93242444 on Build GRCh38
rs755538611 Chr.14: 93242528-93242528 on Build GRCh38
rs145372061 Chr.14: 93242519-93242519 on Build GRCh38
rs767237944 Chr.14: 93242423-93242423 on Build GRCh38
rs552454801 Chr.14: 93242584-93242584 on Build GRCh38
rs755337125 Chr.14: 93242459-93242459 on Build GRCh38
rs779410272 Chr.14: 93242460-93242460 on Build GRCh38
rs145129249 Chr.14: 93242525-93242525 on Build GRCh38
rs61740740 Chr.14: 93242565-93242565 on Build GRCh38
rs748592349 Chr.14: 93242463-93242463 on Build GRCh38
rs572317261 Chr.14: 93242440-93242440 on Build GRCh38
rs201317501 Chr.14: 93242566-93242566 on Build GRCh38
rs139234606 Chr.14: 93242391-93242391 on Build GRCh38
rs772948806 Chr.14: 93242515-93242515 on Build GRCh38
rs771078089 Chr.14: 93242491-93242491 on Build GRCh38
rs763885374 Chr.14: 93242411-93242411 on Build GRCh38
rs141130270 Chr.14: 93242600-93242600 on Build GRCh38
rs200447121 Chr.14: 93242410-93242410 on Build GRCh38
rs904585282 Chr.14: 93242494-93242494 on Build GRCh38
rs878948142 Chr.14: 93242382-93242382 on Build GRCh38
rs1020781822 Chr.14: 93242398-93242398 on Build GRCh38
rs202159154 Chr.14: 93242502-93242502 on Build GRCh38
rs781241929 Chr.14: 93242570-93242570 on Build GRCh38
rs953718840 Chr.14: 93242466-93242466 on Build GRCh38
rs761715181 Chr.14: 93242508-93242508 on Build GRCh38
rs986453546 Chr.14: 93242478-93242478 on Build GRCh38
rs565943949 Chr.14: 93242593-93242593 on Build GRCh38
rs141424765 Chr.14: 93242524-93242524 on Build GRCh38
rs565194914 Chr.14: 93242422-93242422 on Build GRCh38
rs113142695 Chr.14: 93242585-93242585 on Build GRCh38
rs778108156 Chr.14: 93242474-93242474 on Build GRCh38
rs765888349 Chr.14: 93242532-93242532 on Build GRCh38
rs961842817 Chr.14: 93242555-93242555 on Build GRCh38
rs755671512 Chr.14: 93242434-93242434 on Build GRCh38
rs201912928 Chr.14: 93242596-93242596 on Build GRCh38
rs745984819 Chr.14: 93242580-93242580 on Build GRCh38
rs749205895 Chr.14: 93242503-93242503 on Build GRCh38
rs36099812 Chr.14: 93242413-93242413 on Build GRCh38

View Full Product Details