Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44176611-44176852 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB16
Entrez Gene ID
92591
Gene Name
ankyrin repeat and SOCS box containing 16
Gene Aliases
-
UniGene
Hs.534517
Gene Chromosome Location
Chr.17: 44170706-44179083 on Build GRCh38
Transcripts
NM_080863.4 NR_049729.1 NR_049730.1

Gene Symbol
ASB16-AS1
Entrez Gene ID
339201
Gene Name
ASB16 antisense RNA 1
Gene Aliases
C17orf65
UniGene
Hs.656564
Gene Chromosome Location
Chr.17: 44175973-44186717 on Build GRCh38
Transcripts
NM_080863.4 NR_049729.1 NR_049730.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6839428 c.617C>A p.A206E Chr.17: 44176785-44176785 on Build GRCh38
4130130 c.576C>A p.A192A Chr.17: 44176744-44176744 on Build GRCh38
6605532 c.512G>A p.R171H Chr.17: 44176741-44176741 on Build GRCh38
4130132 c.578A>G p.K193R Chr.17: 44176746-44176746 on Build GRCh38
6605527 c.571A>G p.S191G Chr.17: 44176682-44176682 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141744178 Chr.17: 44176770-44176770 on Build GRCh38
rs771825647 Chr.17: 44176648-44176684 on Build GRCh38
rs775181279 Chr.17: 44176706-44176707 on Build GRCh38
rs775059036 Chr.17: 44176698-44176698 on Build GRCh38
rs759755472 Chr.17: 44176806-44176806 on Build GRCh38
rs912280763 Chr.17: 44176823-44176823 on Build GRCh38
rs375066415 Chr.17: 44176789-44176789 on Build GRCh38
rs754357179 Chr.17: 44176642-44176642 on Build GRCh38
rs745783423 Chr.17: 44176687-44176687 on Build GRCh38
rs770668467 Chr.17: 44176680-44176680 on Build GRCh38
rs915045693 Chr.17: 44176833-44176833 on Build GRCh38
rs752781496 Chr.17: 44176666-44176666 on Build GRCh38
rs747784592 Chr.17: 44176794-44176794 on Build GRCh38
rs750670230 Chr.17: 44176826-44176826 on Build GRCh38
rs764133179 Chr.17: 44176817-44176817 on Build GRCh38
rs1046889952 Chr.17: 44176665-44176665 on Build GRCh38
rs781000390 Chr.17: 44176683-44176683 on Build GRCh38
rs763385351 Chr.17: 44176830-44176831 on Build GRCh38
rs760373593 Chr.17: 44176741-44176742 on Build GRCh38
rs201053641 Chr.17: 44176793-44176793 on Build GRCh38
rs754308051 Chr.17: 44176749-44176749 on Build GRCh38
rs768108190 Chr.17: 44176804-44176805 on Build GRCh38
rs75600674 Chr.17: 44176727-44176727 on Build GRCh38
rs1048514470 Chr.17: 44176718-44176718 on Build GRCh38
rs769677740 Chr.17: 44176694-44176694 on Build GRCh38
rs765734522 Chr.17: 44176754-44176754 on Build GRCh38
rs767657892 Chr.17: 44176825-44176825 on Build GRCh38
rs963786176 Chr.17: 44176829-44176829 on Build GRCh38
rs755941532 Chr.17: 44176787-44176787 on Build GRCh38
rs147733989 Chr.17: 44176816-44176816 on Build GRCh38
rs776152473 Chr.17: 44176823-44176824 on Build GRCh38
rs758645271 Chr.17: 44176671-44176671 on Build GRCh38
rs957615506 Chr.17: 44176771-44176771 on Build GRCh38
rs886905122 Chr.17: 44176682-44176682 on Build GRCh38
rs931183316 Chr.17: 44176664-44176664 on Build GRCh38
rs771912610 Chr.17: 44176795-44176795 on Build GRCh38
rs145786587 Chr.17: 44176766-44176766 on Build GRCh38
rs745714756 Chr.17: 44176786-44176786 on Build GRCh38
rs779799451 Chr.17: 44176788-44176788 on Build GRCh38
rs766820301 Chr.17: 44176746-44176746 on Build GRCh38
rs761589553 Chr.17: 44176741-44176741 on Build GRCh38
rs755817222 Chr.17: 44176827-44176827 on Build GRCh38
rs747251981 Chr.17: 44176674-44176674 on Build GRCh38
rs751695008 Chr.17: 44176820-44176820 on Build GRCh38
rs376814407 Chr.17: 44176810-44176810 on Build GRCh38
rs753254670 Chr.17: 44176757-44176757 on Build GRCh38
rs368182961 Chr.17: 44176726-44176726 on Build GRCh38
rs75036136 Chr.17: 44176821-44176821 on Build GRCh38
rs766873502 Chr.17: 44176635-44176635 on Build GRCh38
rs762301933 Chr.17: 44176722-44176722 on Build GRCh38
rs760078029 Chr.17: 44176753-44176753 on Build GRCh38
rs768225675 Chr.17: 44176672-44176672 on Build GRCh38
rs7220138 Chr.17: 44176643-44176643 on Build GRCh38
rs779031857 Chr.17: 44176659-44176659 on Build GRCh38
rs758357718 Chr.17: 44176764-44176764 on Build GRCh38
rs959280906 Chr.17: 44176814-44176814 on Build GRCh38
rs146134954 Chr.17: 44176797-44176797 on Build GRCh38

View Full Product Details