Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 33088869-33089142 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPIFB4
Entrez Gene ID
149954
Gene Name
BPI fold containing family B member 4
Gene Aliases
C20orf186, LPLUNC4, RY2G5, dJ726C3.5
UniGene
Hs.38961
Gene Chromosome Location
Chr.20: 33081512-33111751 on Build GRCh38
Transcripts
NM_182519.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4097644 c.810G>A p.G270G Chr.20: 33088966-33088966 on Build GRCh38
1025873 c.825C>T p.L275L Chr.20: 33088981-33088981 on Build GRCh38
4519098 c.849_850CC>TT p.(=) Chr.20: 33089005-33089006 on Build GRCh38
4001878 c.842A>C p.N281T Chr.20: 33088998-33088998 on Build GRCh38
6612372 c.859G>A p.V287I Chr.20: 33089015-33089015 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1051516484 Chr.20: 33089111-33089111 on Build GRCh38
rs779403776 Chr.20: 33089015-33089015 on Build GRCh38
rs772677932 Chr.20: 33088970-33088970 on Build GRCh38
rs773431276 Chr.20: 33088975-33088975 on Build GRCh38
rs543633125 Chr.20: 33088977-33088977 on Build GRCh38
rs780767139 Chr.20: 33089011-33089011 on Build GRCh38
rs752595746 Chr.20: 33088932-33088932 on Build GRCh38
rs1020518754 Chr.20: 33089109-33089109 on Build GRCh38
rs757051950 Chr.20: 33089008-33089008 on Build GRCh38
rs1027407879 Chr.20: 33088930-33088930 on Build GRCh38
rs764150448 Chr.20: 33089001-33089001 on Build GRCh38
rs769308715 Chr.20: 33088946-33088946 on Build GRCh38
rs1019026273 Chr.20: 33089114-33089114 on Build GRCh38
rs149304342 Chr.20: 33088966-33088966 on Build GRCh38
rs6119321 Chr.20: 33089063-33089063 on Build GRCh38
rs759689350 Chr.20: 33088924-33088924 on Build GRCh38
rs756889167 Chr.20: 33088936-33088936 on Build GRCh38
rs935456256 Chr.20: 33089104-33089104 on Build GRCh38
rs769461492 Chr.20: 33088904-33088904 on Build GRCh38
rs1009198332 Chr.20: 33089121-33089121 on Build GRCh38
rs371608315 Chr.20: 33089000-33089000 on Build GRCh38
rs780890005 Chr.20: 33088942-33088942 on Build GRCh38
rs184859093 Chr.20: 33089097-33089097 on Build GRCh38
rs759638197 Chr.20: 33088982-33088982 on Build GRCh38
rs41289876 Chr.20: 33089094-33089094 on Build GRCh38
rs755583277 Chr.20: 33089014-33089014 on Build GRCh38
rs748881445 Chr.20: 33089037-33089037 on Build GRCh38
rs139268887 Chr.20: 33088994-33088994 on Build GRCh38
rs777001829 Chr.20: 33088981-33088981 on Build GRCh38
rs576611906 Chr.20: 33088902-33088902 on Build GRCh38
rs778344773 Chr.20: 33089058-33089058 on Build GRCh38
rs758147359 Chr.20: 33088957-33088957 on Build GRCh38
rs979200392 Chr.20: 33088917-33088926 on Build GRCh38
rs751419069 Chr.20: 33088934-33088934 on Build GRCh38
rs2889732 Chr.20: 33088998-33088998 on Build GRCh38
rs181836133 Chr.20: 33089012-33089012 on Build GRCh38
rs751351191 Chr.20: 33089007-33089007 on Build GRCh38
rs771442300 Chr.20: 33089074-33089074 on Build GRCh38
rs376059412 Chr.20: 33088933-33088933 on Build GRCh38
rs765260923 Chr.20: 33088986-33088986 on Build GRCh38
rs779428406 Chr.20: 33088953-33088953 on Build GRCh38
rs1001985155 Chr.20: 33089051-33089051 on Build GRCh38
rs758821968 Chr.20: 33089042-33089042 on Build GRCh38
rs111740143 Chr.20: 33089113-33089113 on Build GRCh38
rs761152133 Chr.20: 33089005-33089005 on Build GRCh38
rs765161328 Chr.20: 33088931-33088931 on Build GRCh38
rs994564103 Chr.20: 33088909-33088909 on Build GRCh38
rs966380692 Chr.20: 33088905-33088905 on Build GRCh38
rs144538484 Chr.20: 33088979-33088979 on Build GRCh38

View Full Product Details