Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 75410787-75411060 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMIM21
Entrez Gene ID
284274
Gene Name
small integral membrane protein 21
Gene Aliases
C18orf62
UniGene
Hs.666744
Gene Chromosome Location
Chr.18: 75409476-75455653 on Build GRCh38
Transcripts
NM_001037331.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1680430 c.276G>T p.K92N Chr.18: 75410894-75410894 on Build GRCh38
6274704 c.261-7C>A p.? Chr.18: 75410916-75410916 on Build GRCh38
1737598 c.269G>A p.R90H Chr.18: 75410901-75410901 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs148196575 Chr.18: 75410901-75410901 on Build GRCh38
rs143010711 Chr.18: 75410883-75410883 on Build GRCh38
rs878938290 Chr.18: 75410963-75410963 on Build GRCh38
rs930675238 Chr.18: 75411009-75411009 on Build GRCh38
rs749312633 Chr.18: 75410943-75410943 on Build GRCh38
rs929051066 Chr.18: 75411010-75411010 on Build GRCh38
rs768870411 Chr.18: 75410942-75410943 on Build GRCh38
rs113904028 Chr.18: 75411024-75411024 on Build GRCh38
rs374400213 Chr.18: 75410860-75410860 on Build GRCh38
rs770050906 Chr.18: 75410930-75410930 on Build GRCh38
rs199809461 Chr.18: 75410842-75410842 on Build GRCh38
rs774337630 Chr.18: 75410854-75410854 on Build GRCh38
rs769605964 Chr.18: 75410827-75410827 on Build GRCh38
rs753155521 Chr.18: 75410892-75410892 on Build GRCh38
rs530050293 Chr.18: 75410869-75410869 on Build GRCh38
rs941681425 Chr.18: 75411026-75411026 on Build GRCh38
rs923149376 Chr.18: 75411006-75411006 on Build GRCh38
rs533949074 Chr.18: 75411028-75411028 on Build GRCh38
rs766992465 Chr.18: 75410873-75410873 on Build GRCh38
rs775215805 Chr.18: 75410831-75410831 on Build GRCh38
rs374864997 Chr.18: 75410937-75410937 on Build GRCh38
rs755781025 Chr.18: 75410877-75410877 on Build GRCh38
rs572506925 Chr.18: 75410917-75410917 on Build GRCh38
rs761772469 Chr.18: 75410952-75410952 on Build GRCh38
rs552593498 Chr.18: 75410902-75410902 on Build GRCh38
rs76331965 Chr.18: 75411017-75411017 on Build GRCh38
rs200982937 Chr.18: 75410946-75410946 on Build GRCh38
rs771506878 Chr.18: 75410957-75410957 on Build GRCh38
rs368499895 Chr.18: 75410903-75410903 on Build GRCh38
rs774387594 Chr.18: 75410950-75410953 on Build GRCh38
rs531476883 Chr.18: 75411000-75411000 on Build GRCh38
rs780205185 Chr.18: 75410942-75410942 on Build GRCh38
rs533312235 Chr.18: 75411028-75411028 on Build GRCh38
rs781207276 Chr.18: 75410925-75410925 on Build GRCh38
rs775657795 Chr.18: 75410931-75410931 on Build GRCh38
rs749144555 Chr.18: 75410932-75410932 on Build GRCh38
rs769838960 Chr.18: 75410930-75410930 on Build GRCh38
rs367602336 Chr.18: 75410958-75410958 on Build GRCh38
rs188728173 Chr.18: 75410933-75410933 on Build GRCh38
rs767025352 Chr.18: 75410980-75410980 on Build GRCh38
rs370971309 Chr.18: 75410853-75410853 on Build GRCh38
rs74927953 Chr.18: 75411027-75411028 on Build GRCh38
rs1044467885 Chr.18: 75411027-75411027 on Build GRCh38
rs745696258 Chr.18: 75410913-75410914 on Build GRCh38
rs374633876 Chr.18: 75410975-75410975 on Build GRCh38
rs953974108 Chr.18: 75410861-75410861 on Build GRCh38
rs977902084 Chr.18: 75410921-75410921 on Build GRCh38
rs371557964 Chr.18: 75410915-75410915 on Build GRCh38
rs750133992 Chr.18: 75410875-75410875 on Build GRCh38
rs573193645 Chr.18: 75410818-75410818 on Build GRCh38

View Full Product Details