Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 49709026-49709287 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRISP2
Entrez Gene ID
7180
Gene Name
cysteine rich secretory protein 2
Gene Aliases
CRISP-2, CT36, GAPDL5, TPX1, TSP1
UniGene
Hs.2042
Gene Chromosome Location
Chr.6: 49676665-49714198 on Build GRCh38
Transcripts
NM_001142407.2 NM_001142408.2 NM_001142417.2 NM_001142435.2 NM_001261822.1 NM_003296.3 XM_005249349.1 XM_005249350.1 XM_005249351.3 XM_005249352.3 XM_005249353.3 XM_005249356.1 XM_011514841.1 XM_011514842.1 XM_011514843.2 XM_011514844.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1445011 c.56C>T p.A19V Chr.6: 49709141-49709141 on Build GRCh38
333048 c.21G>T p.L7F Chr.6: 49709176-49709176 on Build GRCh38
742671 c.30T>A p.V10V Chr.6: 49709167-49709167 on Build GRCh38
4397404 c.64A>T p.K22* Chr.6: 49709133-49709133 on Build GRCh38
5405512 c.64A>C p.K22Q Chr.6: 49709133-49709133 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs191043224 Chr.6: 49709056-49709056 on Build GRCh38
rs774698917 Chr.6: 49709183-49709183 on Build GRCh38
rs764766322 Chr.6: 49709129-49709129 on Build GRCh38
rs375015830 Chr.6: 49709160-49709160 on Build GRCh38
rs775796814 Chr.6: 49709198-49709198 on Build GRCh38
rs762460419 Chr.6: 49709228-49709228 on Build GRCh38
rs771147466 Chr.6: 49709081-49709081 on Build GRCh38
rs776728201 Chr.6: 49709055-49709055 on Build GRCh38
rs114507483 Chr.6: 49709125-49709125 on Build GRCh38
rs931707744 Chr.6: 49709052-49709052 on Build GRCh38
rs761201340 Chr.6: 49709210-49709210 on Build GRCh38
rs142085951 Chr.6: 49709182-49709182 on Build GRCh38
rs866144300 Chr.6: 49709202-49709202 on Build GRCh38
rs764533854 Chr.6: 49709220-49709220 on Build GRCh38
rs148635588 Chr.6: 49709179-49709179 on Build GRCh38
rs754870765 Chr.6: 49709163-49709163 on Build GRCh38
rs758006364 Chr.6: 49709146-49709146 on Build GRCh38
rs150713364 Chr.6: 49709195-49709195 on Build GRCh38
rs1020882699 Chr.6: 49709123-49709123 on Build GRCh38
rs187722545 Chr.6: 49709054-49709054 on Build GRCh38
rs199844467 Chr.6: 49709147-49709147 on Build GRCh38
rs774603689 Chr.6: 49709088-49709088 on Build GRCh38
rs749895432 Chr.6: 49709143-49709143 on Build GRCh38
rs767766004 Chr.6: 49709101-49709101 on Build GRCh38
rs973347742 Chr.6: 49709245-49709245 on Build GRCh38
rs767270333 Chr.6: 49709232-49709232 on Build GRCh38
rs73439865 Chr.6: 49709241-49709241 on Build GRCh38
rs73439862 Chr.6: 49709197-49709197 on Build GRCh38
rs748146747 Chr.6: 49709169-49709169 on Build GRCh38
rs868679273 Chr.6: 49709116-49709116 on Build GRCh38
rs781121368 Chr.6: 49709165-49709165 on Build GRCh38
rs368702279 Chr.6: 49709095-49709095 on Build GRCh38
rs958720919 Chr.6: 49709171-49709171 on Build GRCh38
rs555924769 Chr.6: 49709226-49709226 on Build GRCh38
rs760561522 Chr.6: 49709172-49709172 on Build GRCh38

View Full Product Details