Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 23516237-23516504 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C18orf8
Entrez Gene ID
29919
Gene Name
chromosome 18 open reading frame 8
Gene Aliases
HsT2591, MIC1, Mic-1
UniGene
Hs.464779
Gene Chromosome Location
Chr.18: 23503470-23531807 on Build GRCh38
Transcripts
NM_001276342.1 NM_001318707.1 NM_001318708.1 NM_001318709.1 NM_013326.4 XM_011525939.1 XM_017025717.1 XM_005258277.1 XM_005258279.2 XM_011526015.1

Gene Symbol
NPC1
Entrez Gene ID
4864
Gene Name
NPC intracellular cholesterol transporter 1
Gene Aliases
NPC
UniGene
Hs.464779
Gene Chromosome Location
Chr.18: 23506184-23586617 on Build GRCh38
Transcripts
NM_001276342.1 NM_001318707.1 NM_001318708.1 NM_001318709.1 NM_013326.4 XM_011525939.1 XM_017025717.1 XM_005258277.1 XM_005258279.2 XM_011526015.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1388015 c.564G>A p.S188S Chr.18: 23516334-23516334 on Build GRCh38
6858064 c.628G>A p.E210K Chr.18: 23516398-23516398 on Build GRCh38
1733741 c.634G>C p.D212H Chr.18: 23516404-23516404 on Build GRCh38
2885416 c.643A>G p.M215V Chr.18: 23516413-23516413 on Build GRCh38
3422091 c.600G>A p.A200A Chr.18: 23516370-23516370 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774876794 Chr.18: 23516445-23516445 on Build GRCh38
rs533549598 Chr.18: 23516357-23516357 on Build GRCh38
rs374149112 Chr.18: 23516369-23516369 on Build GRCh38
rs754722071 Chr.18: 23516279-23516279 on Build GRCh38
rs200066647 Chr.18: 23516272-23516272 on Build GRCh38
rs1035895463 Chr.18: 23516453-23516453 on Build GRCh38
rs752690577 Chr.18: 23516397-23516397 on Build GRCh38
rs780745817 Chr.18: 23516412-23516412 on Build GRCh38
rs758328006 Chr.18: 23516403-23516403 on Build GRCh38
rs748049630 Chr.18: 23516307-23516307 on Build GRCh38
rs780557357 Chr.18: 23516431-23516431 on Build GRCh38
rs143371359 Chr.18: 23516342-23516342 on Build GRCh38
rs770250549 Chr.18: 23516356-23516356 on Build GRCh38
rs777135810 Chr.18: 23516323-23516323 on Build GRCh38
rs772163414 Chr.18: 23516473-23516473 on Build GRCh38
rs185178985 Chr.18: 23516432-23516432 on Build GRCh38
rs148477145 Chr.18: 23516413-23516413 on Build GRCh38
rs201270524 Chr.18: 23516387-23516387 on Build GRCh38
rs757954049 Chr.18: 23516308-23516308 on Build GRCh38
rs368667300 Chr.18: 23516410-23516410 on Build GRCh38
rs146281079 Chr.18: 23516366-23516366 on Build GRCh38
rs768919670 Chr.18: 23516364-23516364 on Build GRCh38
rs553858189 Chr.18: 23516363-23516363 on Build GRCh38
rs761833969 Chr.18: 23516447-23516447 on Build GRCh38
rs145956918 Chr.18: 23516324-23516324 on Build GRCh38
rs978826640 Chr.18: 23516423-23516423 on Build GRCh38
rs750621524 Chr.18: 23516370-23516370 on Build GRCh38
rs201621222 Chr.18: 23516417-23516417 on Build GRCh38
rs777708274 Chr.18: 23516407-23516407 on Build GRCh38
rs780846585 Chr.18: 23516333-23516333 on Build GRCh38
rs967429153 Chr.18: 23516430-23516430 on Build GRCh38
rs373829222 Chr.18: 23516426-23516426 on Build GRCh38
rs139940827 Chr.18: 23516327-23516327 on Build GRCh38
rs868520116 Chr.18: 23516428-23516428 on Build GRCh38
rs753591927 Chr.18: 23516277-23516277 on Build GRCh38
rs759368123 Chr.18: 23516372-23516372 on Build GRCh38
rs575941945 Chr.18: 23516440-23516440 on Build GRCh38

View Full Product Details