Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 186197438-186197699 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP4V2
Entrez Gene ID
285440
Gene Name
cytochrome P450 family 4 subfamily V member 2
Gene Aliases
BCD, CYP4AH1
UniGene
Hs.587231
Gene Chromosome Location
Chr.4: 186191520-186213463 on Build GRCh38
Transcripts
NM_207352.3 XM_005262935.3 XM_017008037.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
733878 c.606A>T p.E202D Chr.4: 186197534-186197534 on Build GRCh38
6438813 c.664G>A p.A222T Chr.4: 186197592-186197592 on Build GRCh38
5805270 c.605-2A>T p.? Chr.4: 186197531-186197531 on Build GRCh38
5925094 c.605-1G>A p.? Chr.4: 186197532-186197532 on Build GRCh38
6259010 c.638G>T p.S213I Chr.4: 186197566-186197566 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199476191 Chr.4: 186197583-186197583 on Build GRCh38
rs368178580 Chr.4: 186197512-186197512 on Build GRCh38
rs779063197 Chr.4: 186197529-186197529 on Build GRCh38
rs1035486249 Chr.4: 186197636-186197636 on Build GRCh38
rs201205689 Chr.4: 186197616-186197616 on Build GRCh38
rs765384088 Chr.4: 186197485-186197485 on Build GRCh38
rs757247567 Chr.4: 186197527-186197527 on Build GRCh38
rs4862662 Chr.4: 186197508-186197508 on Build GRCh38
rs372884044 Chr.4: 186197648-186197648 on Build GRCh38
rs200664958 Chr.4: 186197570-186197570 on Build GRCh38
rs763083895 Chr.4: 186197590-186197590 on Build GRCh38
rs930980339 Chr.4: 186197649-186197649 on Build GRCh38
rs748856336 Chr.4: 186197520-186197520 on Build GRCh38
rs770048736 Chr.4: 186197580-186197580 on Build GRCh38
rs752414489 Chr.4: 186197501-186197501 on Build GRCh38
rs562165079 Chr.4: 186197480-186197480 on Build GRCh38
rs755771549 Chr.4: 186197654-186197654 on Build GRCh38
rs773317728 Chr.4: 186197490-186197490 on Build GRCh38
rs372015953 Chr.4: 186197517-186197517 on Build GRCh38
rs751913598 Chr.4: 186197494-186197494 on Build GRCh38
rs972876509 Chr.4: 186197633-186197633 on Build GRCh38
rs1000127688 Chr.4: 186197511-186197511 on Build GRCh38
rs367800547 Chr.4: 186197495-186197495 on Build GRCh38
rs766803737 Chr.4: 186197607-186197607 on Build GRCh38
rs761744214 Chr.4: 186197483-186197483 on Build GRCh38
rs376640607 Chr.4: 186197554-186197554 on Build GRCh38
rs763425913 Chr.4: 186197491-186197491 on Build GRCh38
rs190069196 Chr.4: 186197482-186197482 on Build GRCh38
rs745471184 Chr.4: 186197589-186197589 on Build GRCh38
rs767954060 Chr.4: 186197644-186197644 on Build GRCh38
rs771363802 Chr.4: 186197639-186197642 on Build GRCh38
rs34331648 Chr.4: 186197566-186197566 on Build GRCh38
rs766926054 Chr.4: 186197492-186197492 on Build GRCh38
rs749762667 Chr.4: 186197483-186197484 on Build GRCh38
rs61745524 Chr.4: 186197538-186197538 on Build GRCh38
rs372020737 Chr.4: 186197496-186197496 on Build GRCh38
rs548721610 Chr.4: 186197464-186197464 on Build GRCh38
rs187025073 Chr.4: 186197613-186197613 on Build GRCh38

View Full Product Details