Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42683995-42684268 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTNAP1
Entrez Gene ID
8506
Gene Name
contactin associated protein 1
Gene Aliases
CASPR, CNTNAP, NRXN4, P190
UniGene
Hs.408730
Gene Chromosome Location
Chr.17: 42679963-42699993 on Build GRCh38
Transcripts
NM_003632.2 XM_005257748.4 XM_017025238.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4623560 c.295C>T p.R99C Chr.17: 42684161-42684161 on Build GRCh38
1201690 c.170-2A>G p.? Chr.17: 42684034-42684034 on Build GRCh38
6002551 c.178G>A p.G60R Chr.17: 42684044-42684044 on Build GRCh38
1710315 c.191_192GG>AA p.R64Q Chr.17: 42684057-42684058 on Build GRCh38
472831 c.359A>G p.N120S Chr.17: 42684225-42684225 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs115552583 Chr.17: 42684193-42684193 on Build GRCh38
rs144497813 Chr.17: 42684096-42684096 on Build GRCh38
rs771973268 Chr.17: 42684243-42684243 on Build GRCh38
rs745452887 Chr.17: 42684050-42684050 on Build GRCh38
rs768684553 Chr.17: 42684110-42684110 on Build GRCh38
rs748839835 Chr.17: 42684209-42684209 on Build GRCh38
rs373439152 Chr.17: 42684033-42684033 on Build GRCh38
rs369648544 Chr.17: 42684030-42684030 on Build GRCh38
rs764615609 Chr.17: 42684166-42684166 on Build GRCh38
rs767706478 Chr.17: 42684179-42684179 on Build GRCh38
rs114871554 Chr.17: 42684244-42684244 on Build GRCh38
rs530020899 Chr.17: 42684114-42684114 on Build GRCh38
rs778155999 Chr.17: 42684027-42684027 on Build GRCh38
rs779751302 Chr.17: 42684204-42684204 on Build GRCh38
rs1012582830 Chr.17: 42684082-42684082 on Build GRCh38
rs901717903 Chr.17: 42684178-42684178 on Build GRCh38
rs376723451 Chr.17: 42684016-42684016 on Build GRCh38
rs375684503 Chr.17: 42684228-42684228 on Build GRCh38
rs765414012 Chr.17: 42684138-42684138 on Build GRCh38
rs775520960 Chr.17: 42684155-42684155 on Build GRCh38
rs1019694046 Chr.17: 42684177-42684177 on Build GRCh38
rs755718283 Chr.17: 42684055-42684055 on Build GRCh38
rs747983304 Chr.17: 42684118-42684118 on Build GRCh38
rs780917967 Chr.17: 42684038-42684038 on Build GRCh38
rs752012820 Chr.17: 42684175-42684175 on Build GRCh38
rs779011841 Chr.17: 42684222-42684222 on Build GRCh38
rs771709079 Chr.17: 42684119-42684119 on Build GRCh38
rs776920122 Chr.17: 42684132-42684132 on Build GRCh38
rs749287940 Chr.17: 42684107-42684107 on Build GRCh38
rs759670106 Chr.17: 42684135-42684135 on Build GRCh38
rs1008347706 Chr.17: 42684133-42684133 on Build GRCh38
rs370354894 Chr.17: 42684181-42684181 on Build GRCh38
rs201128063 Chr.17: 42684240-42684240 on Build GRCh38
rs754492275 Chr.17: 42684219-42684219 on Build GRCh38
rs772210623 Chr.17: 42684044-42684044 on Build GRCh38

View Full Product Details