Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 143867053-143867302 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPPK1
Entrez Gene ID
83481
Gene Name
epiplakin 1
Gene Aliases
EPIPL, EPIPL1
UniGene
Hs.200412
Gene Chromosome Location
Chr.8: 143857319-143879043 on Build GRCh38
Transcripts
NM_031308.3 XM_017013889.1 XM_017013890.1 XM_017013891.1 XM_017013892.1

SNP Detail

SNP ID Chromosome Location
rs782149613 Chr.8: 143867279-143867279 on Build GRCh38
rs375002026 Chr.8: 143867214-143867214 on Build GRCh38
rs781861594 Chr.8: 143867209-143867210 on Build GRCh38
rs373165682 Chr.8: 143867260-143867260 on Build GRCh38
rs35176503 Chr.8: 143867224-143867224 on Build GRCh38
rs781893307 Chr.8: 143867100-143867100 on Build GRCh38
rs528153180 Chr.8: 143867259-143867259 on Build GRCh38
rs782245728 Chr.8: 143867122-143867122 on Build GRCh38
rs782379600 Chr.8: 143867174-143867174 on Build GRCh38
rs782691764 Chr.8: 143867261-143867261 on Build GRCh38
rs782195794 Chr.8: 143867211-143867211 on Build GRCh38
rs553910552 Chr.8: 143867078-143867078 on Build GRCh38
rs782820648 Chr.8: 143867132-143867132 on Build GRCh38
rs554910033 Chr.8: 143867083-143867083 on Build GRCh38
rs782819737 Chr.8: 143867082-143867082 on Build GRCh38
rs139977710 Chr.8: 143867202-143867202 on Build GRCh38
rs782711807 Chr.8: 143867097-143867097 on Build GRCh38
rs977353069 Chr.8: 143867123-143867123 on Build GRCh38
rs782724618 Chr.8: 143867136-143867136 on Build GRCh38
rs781847135 Chr.8: 143867133-143867133 on Build GRCh38
rs782461729 Chr.8: 143867199-143867199 on Build GRCh38
rs368863679 Chr.8: 143867131-143867131 on Build GRCh38
rs782373505 Chr.8: 143867263-143867263 on Build GRCh38
rs781951419 Chr.8: 143867266-143867266 on Build GRCh38
rs527896611 Chr.8: 143867156-143867156 on Build GRCh38
rs189688618 Chr.8: 143867157-143867157 on Build GRCh38
rs782768826 Chr.8: 143867244-143867244 on Build GRCh38
rs782182155 Chr.8: 143867115-143867115 on Build GRCh38
rs545639287 Chr.8: 143867220-143867220 on Build GRCh38
rs782322639 Chr.8: 143867117-143867117 on Build GRCh38
rs782230626 Chr.8: 143867272-143867272 on Build GRCh38
rs782653009 Chr.8: 143867204-143867204 on Build GRCh38
rs782150025 Chr.8: 143867183-143867183 on Build GRCh38
rs199828972 Chr.8: 143867280-143867280 on Build GRCh38
rs782207878 Chr.8: 143867153-143867153 on Build GRCh38
rs782200809 Chr.8: 143867208-143867208 on Build GRCh38
rs782734320 Chr.8: 143867223-143867223 on Build GRCh38
rs781865685 Chr.8: 143867246-143867246 on Build GRCh38
rs781793059 Chr.8: 143867193-143867193 on Build GRCh38
rs146515033 Chr.8: 143867144-143867144 on Build GRCh38
rs782107684 Chr.8: 143867172-143867172 on Build GRCh38
rs572291032 Chr.8: 143867079-143867079 on Build GRCh38
rs782662791 Chr.8: 143867152-143867152 on Build GRCh38
rs571579347 Chr.8: 143867216-143867216 on Build GRCh38
rs782042148 Chr.8: 143867170-143867170 on Build GRCh38
rs782809770 Chr.8: 143867184-143867184 on Build GRCh38
rs375707682 Chr.8: 143867129-143867129 on Build GRCh38
rs78008227 Chr.8: 143867191-143867191 on Build GRCh38
rs781845272 Chr.8: 143867090-143867090 on Build GRCh38
rs782481607 Chr.8: 143867187-143867187 on Build GRCh38
rs782390496 Chr.8: 143867125-143867125 on Build GRCh38
rs186320706 Chr.8: 143867118-143867118 on Build GRCh38
rs1020981879 Chr.8: 143867257-143867257 on Build GRCh38
rs782125364 Chr.8: 143867134-143867134 on Build GRCh38
rs367640831 Chr.8: 143867252-143867252 on Build GRCh38
rs782561343 Chr.8: 143867235-143867235 on Build GRCh38
rs782089709 Chr.8: 143867178-143867178 on Build GRCh38
rs1036757233 Chr.8: 143867177-143867177 on Build GRCh38
rs182185580 Chr.8: 143867113-143867113 on Build GRCh38
rs12681478 Chr.8: 143867251-143867251 on Build GRCh38
rs536513369 Chr.8: 143867080-143867080 on Build GRCh38
rs575399844 Chr.8: 143867215-143867215 on Build GRCh38
rs782583923 Chr.8: 143867119-143867119 on Build GRCh38
rs781868121 Chr.8: 143867105-143867105 on Build GRCh38
rs782405559 Chr.8: 143867212-143867212 on Build GRCh38
rs782364626 Chr.8: 143867210-143867210 on Build GRCh38
rs782457896 Chr.8: 143867093-143867093 on Build GRCh38
rs372460806 Chr.8: 143867102-143867102 on Build GRCh38
rs782033531 Chr.8: 143867213-143867213 on Build GRCh38
rs540799146 Chr.8: 143867151-143867151 on Build GRCh38
rs781885118 Chr.8: 143867149-143867149 on Build GRCh38
rs376941147 Chr.8: 143867278-143867278 on Build GRCh38
rs782050033 Chr.8: 143867222-143867222 on Build GRCh38
rs560287631 Chr.8: 143867155-143867155 on Build GRCh38
rs782452886 Chr.8: 143867096-143867096 on Build GRCh38
rs375927494 Chr.8: 143867181-143867181 on Build GRCh38
rs781835809 Chr.8: 143867140-143867140 on Build GRCh38
rs782411839 Chr.8: 143867164-143867164 on Build GRCh38
rs782696825 Chr.8: 143867169-143867169 on Build GRCh38
rs782510861 Chr.8: 143867203-143867203 on Build GRCh38
rs782446058 Chr.8: 143867254-143867254 on Build GRCh38

View Full Product Details