Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 32716192-32716441 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNOT10
Entrez Gene ID
25904
Gene Name
CCR4-NOT transcription complex subunit 10
Gene Aliases
-
UniGene
Hs.444851
Gene Chromosome Location
Chr.3: 32685145-32773875 on Build GRCh38
Transcripts
NM_001256741.1 NM_001256742.1 NM_015442.2 NR_046352.1 XM_006713084.3 XM_006713085.3 XM_011533566.2 XM_011533567.2 XM_017006109.1 XM_017006110.1 XM_017006111.1 XM_017006112.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5431006 c.754-6C>G p.? Chr.3: 32716219-32716219 on Build GRCh38
3592461 c.658C>T p.Q220* Chr.3: 32716309-32716309 on Build GRCh38
4935200 c.594A>G p.K198K Chr.3: 32716245-32716245 on Build GRCh38
5431005 c.574-6C>G p.? Chr.3: 32716219-32716219 on Build GRCh38
3592460 c.577G>C p.G193R Chr.3: 32716228-32716228 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs557882901 Chr.3: 32716379-32716379 on Build GRCh38
rs887257032 Chr.3: 32716337-32716337 on Build GRCh38
rs143945309 Chr.3: 32716258-32716258 on Build GRCh38
rs367870896 Chr.3: 32716302-32716302 on Build GRCh38
rs1035059462 Chr.3: 32716246-32716246 on Build GRCh38
rs760760517 Chr.3: 32716317-32716317 on Build GRCh38
rs144176493 Chr.3: 32716304-32716304 on Build GRCh38
rs747346145 Chr.3: 32716344-32716344 on Build GRCh38
rs959065159 Chr.3: 32716287-32716287 on Build GRCh38
rs934534355 Chr.3: 32716249-32716249 on Build GRCh38
rs986862346 Chr.3: 32716377-32716377 on Build GRCh38
rs76955747 Chr.3: 32716405-32716405 on Build GRCh38
rs752978385 Chr.3: 32716315-32716315 on Build GRCh38
rs750155347 Chr.3: 32716326-32716326 on Build GRCh38
rs1054756038 Chr.3: 32716281-32716281 on Build GRCh38
rs374333174 Chr.3: 32716238-32716238 on Build GRCh38
rs751180798 Chr.3: 32716348-32716348 on Build GRCh38
rs983663932 Chr.3: 32716334-32716334 on Build GRCh38
rs372082520 Chr.3: 32716303-32716303 on Build GRCh38
rs142149948 Chr.3: 32716346-32716347 on Build GRCh38
rs763545278 Chr.3: 32716271-32716271 on Build GRCh38
rs765112401 Chr.3: 32716318-32716318 on Build GRCh38
rs201849234 Chr.3: 32716232-32716232 on Build GRCh38
rs902237388 Chr.3: 32716236-32716236 on Build GRCh38
rs773396632 Chr.3: 32716270-32716270 on Build GRCh38
rs535417501 Chr.3: 32716346-32716346 on Build GRCh38
rs780786295 Chr.3: 32716356-32716356 on Build GRCh38
rs144659723 Chr.3: 32716370-32716370 on Build GRCh38
rs774751406 Chr.3: 32716335-32716335 on Build GRCh38
rs771408169 Chr.3: 32716274-32716274 on Build GRCh38
rs758457690 Chr.3: 32716333-32716333 on Build GRCh38
rs780124059 Chr.3: 32716341-32716341 on Build GRCh38
rs547098322 Chr.3: 32716349-32716349 on Build GRCh38
rs201770145 Chr.3: 32716309-32716309 on Build GRCh38
rs1047503167 Chr.3: 32716322-32716322 on Build GRCh38
rs770205168 Chr.3: 32716263-32716263 on Build GRCh38
rs955385957 Chr.3: 32716376-32716376 on Build GRCh38
rs115116954 Chr.3: 32716369-32716369 on Build GRCh38

View Full Product Details