Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 977243-977482 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMRT3
Entrez Gene ID
58524
Gene Name
doublesex and mab-3 related transcription factor 3
Gene Aliases
DMRTA3
UniGene
Hs.189174
Gene Chromosome Location
Chr.9: 976968-991732 on Build GRCh38
Transcripts
NM_021240.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6217731 c.375C>T p.A125A Chr.9: 977376-977376 on Build GRCh38
5467778 c.252G>T p.E84D Chr.9: 977253-977253 on Build GRCh38
6645683 c.275G>A p.R92H Chr.9: 977276-977276 on Build GRCh38
6189399 c.304G>C p.D102H Chr.9: 977305-977305 on Build GRCh38
5450606 c.365G>A p.R122H Chr.9: 977366-977366 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1033535846 Chr.9: 977263-977263 on Build GRCh38
rs764727932 Chr.9: 977270-977270 on Build GRCh38
rs775570635 Chr.9: 977376-977376 on Build GRCh38
rs926164157 Chr.9: 977347-977347 on Build GRCh38
rs573423358 Chr.9: 977397-977397 on Build GRCh38
rs747917599 Chr.9: 977431-977431 on Build GRCh38
rs755840866 Chr.9: 977290-977290 on Build GRCh38
rs769046798 Chr.9: 977310-977312 on Build GRCh38
rs775580577 Chr.9: 977462-977462 on Build GRCh38
rs950153747 Chr.9: 977366-977366 on Build GRCh38
rs368615844 Chr.9: 977274-977274 on Build GRCh38
rs535809646 Chr.9: 977420-977420 on Build GRCh38
rs748909047 Chr.9: 977418-977418 on Build GRCh38
rs772182173 Chr.9: 977436-977436 on Build GRCh38
rs891913305 Chr.9: 977365-977365 on Build GRCh38
rs759662780 Chr.9: 977326-977326 on Build GRCh38
rs758676642 Chr.9: 977269-977269 on Build GRCh38
rs773265105 Chr.9: 977438-977438 on Build GRCh38
rs780713887 Chr.9: 977279-977279 on Build GRCh38
rs774752054 Chr.9: 977311-977324 on Build GRCh38
rs1026486767 Chr.9: 977284-977284 on Build GRCh38
rs745352669 Chr.9: 977281-977281 on Build GRCh38
rs746027182 Chr.9: 977441-977441 on Build GRCh38
rs776887239 Chr.9: 977321-977321 on Build GRCh38
rs749178060 Chr.9: 977301-977301 on Build GRCh38
rs975010275 Chr.9: 977325-977325 on Build GRCh38
rs963558725 Chr.9: 977272-977272 on Build GRCh38
rs757714876 Chr.9: 977275-977275 on Build GRCh38
rs779553116 Chr.9: 977417-977417 on Build GRCh38
rs765553643 Chr.9: 977367-977367 on Build GRCh38
rs757802743 Chr.9: 977400-977400 on Build GRCh38
rs772092390 Chr.9: 977317-977317 on Build GRCh38
rs778879220 Chr.9: 977425-977425 on Build GRCh38
rs769766595 Chr.9: 977454-977454 on Build GRCh38
rs148355218 Chr.9: 977312-977312 on Build GRCh38
rs552693480 Chr.9: 977262-977262 on Build GRCh38
rs763167788 Chr.9: 977391-977391 on Build GRCh38
rs767989863 Chr.9: 977405-977405 on Build GRCh38
rs764377835 Chr.9: 977395-977395 on Build GRCh38
rs1001464207 Chr.9: 977401-977401 on Build GRCh38
rs555748279 Chr.9: 977463-977463 on Build GRCh38
rs371952660 Chr.9: 977295-977295 on Build GRCh38
rs936174469 Chr.9: 977347-977348 on Build GRCh38
rs1053251914 Chr.9: 977359-977359 on Build GRCh38
rs753150226 Chr.9: 977265-977265 on Build GRCh38
rs749993647 Chr.9: 977408-977408 on Build GRCh38
rs755578612 Chr.9: 977410-977410 on Build GRCh38
rs748116341 Chr.9: 977314-977314 on Build GRCh38
rs200227684 Chr.9: 977308-977308 on Build GRCh38
rs1023606643 Chr.9: 977443-977443 on Build GRCh38

View Full Product Details