Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 169753615-169753888 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HPF1
Entrez Gene ID
54969
Gene Name
histone PARylation factor 1
Gene Aliases
C4orf27
UniGene
Hs.406756
Gene Chromosome Location
Chr.4: 169729468-169757954 on Build GRCh38
Transcripts
NM_017867.2 XM_017008332.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3428333 c.112C>T p.R38* Chr.4: 169753772-169753772 on Build GRCh38
1633556 c.58_59insA p.T20fs*3 Chr.4: 169753825-169753826 on Build GRCh38
1618592 c.164A>G p.H55R Chr.4: 169753720-169753720 on Build GRCh38
7235959 c.91G>T p.A31S Chr.4: 169753793-169753793 on Build GRCh38
6480479 c.134A>G p.Y45C Chr.4: 169753750-169753750 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776600315 Chr.4: 169753799-169753799 on Build GRCh38
rs751758031 Chr.4: 169753662-169753662 on Build GRCh38
rs116110133 Chr.4: 169753658-169753658 on Build GRCh38
rs199937559 Chr.4: 169753844-169753844 on Build GRCh38
rs780445157 Chr.4: 169753846-169753846 on Build GRCh38
rs761705195 Chr.4: 169753815-169753815 on Build GRCh38
rs775297235 Chr.4: 169753786-169753786 on Build GRCh38
rs764035101 Chr.4: 169753652-169753652 on Build GRCh38
rs759615106 Chr.4: 169753847-169753847 on Build GRCh38
rs755003026 Chr.4: 169753793-169753806 on Build GRCh38
rs111514534 Chr.4: 169753661-169753661 on Build GRCh38
rs749803371 Chr.4: 169753752-169753752 on Build GRCh38
rs553719189 Chr.4: 169753758-169753758 on Build GRCh38
rs745405292 Chr.4: 169753767-169753767 on Build GRCh38
rs182953910 Chr.4: 169753855-169753855 on Build GRCh38
rs555565640 Chr.4: 169753849-169753849 on Build GRCh38
rs910794920 Chr.4: 169753858-169753858 on Build GRCh38
rs755142162 Chr.4: 169753666-169753666 on Build GRCh38
rs756602299 Chr.4: 169753660-169753660 on Build GRCh38
rs775387327 Chr.4: 169753644-169753644 on Build GRCh38
rs778521095 Chr.4: 169753766-169753766 on Build GRCh38
rs375267330 Chr.4: 169753645-169753645 on Build GRCh38
rs765180569 Chr.4: 169753820-169753820 on Build GRCh38
rs544388773 Chr.4: 169753860-169753860 on Build GRCh38
rs770456565 Chr.4: 169753762-169753762 on Build GRCh38
rs565649683 Chr.4: 169753772-169753772 on Build GRCh38
rs770744830 Chr.4: 169753723-169753723 on Build GRCh38
rs574383841 Chr.4: 169753741-169753741 on Build GRCh38
rs760552529 Chr.4: 169753648-169753648 on Build GRCh38
rs756445516 Chr.4: 169753733-169753733 on Build GRCh38
rs756492279 Chr.4: 169753856-169753856 on Build GRCh38
rs760506842 Chr.4: 169753788-169753788 on Build GRCh38
rs753778530 Chr.4: 169753655-169753655 on Build GRCh38
rs771875861 Chr.4: 169753643-169753643 on Build GRCh38
rs538302901 Chr.4: 169753691-169753691 on Build GRCh38
rs768402160 Chr.4: 169753798-169753798 on Build GRCh38

View Full Product Details