Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 44484163-44484422 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTDSPL2
Entrez Gene ID
51496
Gene Name
CTD small phosphatase like 2
Gene Aliases
HSPC058, HSPC129
UniGene
Hs.497967
Gene Chromosome Location
Chr.15: 44427329-44527774 on Build GRCh38
Transcripts
NM_016396.2 XM_005254441.2 XM_006720565.2 XM_011521665.2 XM_017022306.1 XM_017022307.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
470699 c.204T>G p.P68P Chr.15: 44484241-44484241 on Build GRCh38
5495629 c.187-10A>G p.? Chr.15: 44484214-44484214 on Build GRCh38
6328791 c.211C>T p.R71W Chr.15: 44484248-44484248 on Build GRCh38
962101 c.212G>A p.R71Q Chr.15: 44484249-44484249 on Build GRCh38
6637411 c.203C>A p.P68H Chr.15: 44484240-44484240 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767349011 Chr.15: 44484212-44484213 on Build GRCh38
rs747007475 Chr.15: 44484286-44484286 on Build GRCh38
rs369801981 Chr.15: 44484288-44484288 on Build GRCh38
rs753067001 Chr.15: 44484358-44484358 on Build GRCh38
rs779941043 Chr.15: 44484335-44484335 on Build GRCh38
rs760992786 Chr.15: 44484373-44484373 on Build GRCh38
rs140283027 Chr.15: 44484321-44484321 on Build GRCh38
rs769574304 Chr.15: 44484317-44484317 on Build GRCh38
rs770519944 Chr.15: 44484326-44484326 on Build GRCh38
rs150517746 Chr.15: 44484264-44484264 on Build GRCh38
rs764969428 Chr.15: 44484208-44484208 on Build GRCh38
rs768546379 Chr.15: 44484195-44484195 on Build GRCh38
rs767957106 Chr.15: 44484333-44484333 on Build GRCh38
rs776537810 Chr.15: 44484205-44484205 on Build GRCh38
rs375511811 Chr.15: 44484279-44484279 on Build GRCh38
rs112342070 Chr.15: 44484236-44484236 on Build GRCh38
rs761626247 Chr.15: 44484206-44484206 on Build GRCh38
rs777907964 Chr.15: 44484275-44484275 on Build GRCh38
rs141681506 Chr.15: 44484253-44484253 on Build GRCh38
rs766825571 Chr.15: 44484214-44484214 on Build GRCh38
rs866787376 Chr.15: 44484295-44484295 on Build GRCh38
rs764325446 Chr.15: 44484382-44484382 on Build GRCh38
rs759411875 Chr.15: 44484208-44484209 on Build GRCh38
rs947548790 Chr.15: 44484329-44484329 on Build GRCh38
rs753052510 Chr.15: 44484266-44484266 on Build GRCh38
rs776627508 Chr.15: 44484290-44484290 on Build GRCh38
rs757364972 Chr.15: 44484269-44484269 on Build GRCh38
rs772958252 Chr.15: 44484319-44484319 on Build GRCh38
rs770722660 Chr.15: 44484200-44484200 on Build GRCh38
rs371305927 Chr.15: 44484263-44484263 on Build GRCh38
rs762458397 Chr.15: 44484211-44484211 on Build GRCh38
rs773991313 Chr.15: 44484199-44484200 on Build GRCh38
rs772859628 Chr.15: 44484209-44484209 on Build GRCh38
rs1053693886 Chr.15: 44484296-44484296 on Build GRCh38
rs1044798616 Chr.15: 44484270-44484270 on Build GRCh38
rs749363455 Chr.15: 44484277-44484277 on Build GRCh38
rs778837136 Chr.15: 44484284-44484284 on Build GRCh38
rs142680502 Chr.15: 44484330-44484330 on Build GRCh38
rs372675259 Chr.15: 44484306-44484306 on Build GRCh38

View Full Product Details