Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 123040577-123040804 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACADSB
Entrez Gene ID
36
Gene Name
acyl-CoA dehydrogenase, short/branched chain
Gene Aliases
2-MEBCAD, ACAD7, SBCAD
UniGene
Hs.81934
Gene Chromosome Location
Chr.10: 123008913-123058290 on Build GRCh38
Transcripts
NM_001609.3 XM_017016194.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4144413 c.444A>T p.T148T Chr.10: 123040606-123040606 on Build GRCh38
4012369 c.479C>T p.A160V Chr.10: 123040641-123040641 on Build GRCh38
915745 c.505G>T p.E169* Chr.10: 123040667-123040667 on Build GRCh38
5560737 c.428A>T p.N143I Chr.10: 123040590-123040590 on Build GRCh38
6591702 c.473delA p.K159fs*13 Chr.10: 123040635-123040635 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140747102 Chr.10: 123040614-123040614 on Build GRCh38
rs760587087 Chr.10: 123040695-123040695 on Build GRCh38
rs754669912 Chr.10: 123040708-123040708 on Build GRCh38
rs369589223 Chr.10: 123040726-123040726 on Build GRCh38
rs767301576 Chr.10: 123040680-123040680 on Build GRCh38
rs773089971 Chr.10: 123040685-123040685 on Build GRCh38
rs769921991 Chr.10: 123040626-123040626 on Build GRCh38
rs567653200 Chr.10: 123040738-123040738 on Build GRCh38
rs150088245 Chr.10: 123040616-123040616 on Build GRCh38
rs756568861 Chr.10: 123040765-123040765 on Build GRCh38
rs766252344 Chr.10: 123040699-123040699 on Build GRCh38
rs546175734 Chr.10: 123040757-123040757 on Build GRCh38
rs775429837 Chr.10: 123040640-123040640 on Build GRCh38
rs186081101 Chr.10: 123040658-123040658 on Build GRCh38
rs369360411 Chr.10: 123040705-123040705 on Build GRCh38
rs761815166 Chr.10: 123040660-123040660 on Build GRCh38
rs764805192 Chr.10: 123040713-123040713 on Build GRCh38
rs1046972266 Chr.10: 123040628-123040628 on Build GRCh38
rs182966982 Chr.10: 123040657-123040657 on Build GRCh38
rs555765069 Chr.10: 123040643-123040643 on Build GRCh38
rs752086697 Chr.10: 123040714-123040714 on Build GRCh38

View Full Product Details