Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 73556161-73556434 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
REC114
Entrez Gene ID
283677
Gene Name
REC114 meiotic recombination protein
Gene Aliases
C15orf60, CT147
UniGene
Hs.743574
Gene Chromosome Location
Chr.15: 73443158-73560015 on Build GRCh38
Transcripts
NM_001042367.1 XM_017022083.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3794390 c.570G>C p.Q190H Chr.15: 73556325-73556325 on Build GRCh38
5032193 c.618A>C p.S206S Chr.15: 73556373-73556373 on Build GRCh38
294186 c.623C>T p.T208M Chr.15: 73556378-73556378 on Build GRCh38
6614466 c.586G>A p.A196T Chr.15: 73556341-73556341 on Build GRCh38
5808057 c.568C>A p.Q190K Chr.15: 73556323-73556323 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750815476 Chr.15: 73556322-73556322 on Build GRCh38
rs372790389 Chr.15: 73556329-73556329 on Build GRCh38
rs900917883 Chr.15: 73556272-73556272 on Build GRCh38
rs776231411 Chr.15: 73556352-73556352 on Build GRCh38
rs770091273 Chr.15: 73556195-73556195 on Build GRCh38
rs371646054 Chr.15: 73556261-73556261 on Build GRCh38
rs766800551 Chr.15: 73556318-73556320 on Build GRCh38
rs201063024 Chr.15: 73556361-73556361 on Build GRCh38
rs371799830 Chr.15: 73556339-73556339 on Build GRCh38
rs756524837 Chr.15: 73556397-73556397 on Build GRCh38
rs547649692 Chr.15: 73556217-73556217 on Build GRCh38
rs1042097935 Chr.15: 73556356-73556356 on Build GRCh38
rs767789564 Chr.15: 73556381-73556381 on Build GRCh38
rs115150158 Chr.15: 73556342-73556342 on Build GRCh38
rs756515141 Chr.15: 73556331-73556331 on Build GRCh38
rs369637394 Chr.15: 73556348-73556348 on Build GRCh38
rs764358509 Chr.15: 73556303-73556303 on Build GRCh38
rs771620981 Chr.15: 73556340-73556340 on Build GRCh38
rs776624996 Chr.15: 73556278-73556278 on Build GRCh38
rs748317896 Chr.15: 73556233-73556233 on Build GRCh38
rs761703087 Chr.15: 73556243-73556243 on Build GRCh38
rs961404961 Chr.15: 73556275-73556275 on Build GRCh38
rs765232982 Chr.15: 73556289-73556289 on Build GRCh38
rs760442984 Chr.15: 73556274-73556274 on Build GRCh38
rs754384456 Chr.15: 73556398-73556398 on Build GRCh38
rs759379156 Chr.15: 73556353-73556353 on Build GRCh38
rs750709030 Chr.15: 73556286-73556288 on Build GRCh38
rs757901000 Chr.15: 73556399-73556399 on Build GRCh38
rs929764546 Chr.15: 73556282-73556282 on Build GRCh38
rs758519881 Chr.15: 73556298-73556298 on Build GRCh38
rs770754178 Chr.15: 73556276-73556276 on Build GRCh38
rs1013831613 Chr.15: 73556383-73556383 on Build GRCh38
rs532484259 Chr.15: 73556216-73556216 on Build GRCh38
rs759447311 Chr.15: 73556284-73556284 on Build GRCh38
rs945045460 Chr.15: 73556350-73556350 on Build GRCh38
rs765219966 Chr.15: 73556274-73556276 on Build GRCh38
rs1014276171 Chr.15: 73556244-73556244 on Build GRCh38
rs376531219 Chr.15: 73556364-73556364 on Build GRCh38
rs911365916 Chr.15: 73556209-73556209 on Build GRCh38
rs368274140 Chr.15: 73556305-73556305 on Build GRCh38
rs373443142 Chr.15: 73556362-73556362 on Build GRCh38
rs200297290 Chr.15: 73556335-73556335 on Build GRCh38
rs376170807 Chr.15: 73556271-73556271 on Build GRCh38
rs750717738 Chr.15: 73556391-73556391 on Build GRCh38
rs778232223 Chr.15: 73556255-73556255 on Build GRCh38
rs112737749 Chr.15: 73556379-73556379 on Build GRCh38
rs369531036 Chr.15: 73556343-73556343 on Build GRCh38
rs752652740 Chr.15: 73556293-73556293 on Build GRCh38
rs757662196 Chr.15: 73556307-73556307 on Build GRCh38
rs751926956 Chr.15: 73556383-73556383 on Build GRCh38
rs781470434 Chr.15: 73556320-73556320 on Build GRCh38
rs771673836 Chr.15: 73556262-73556262 on Build GRCh38
rs777321480 Chr.15: 73556401-73556401 on Build GRCh38
rs751821114 Chr.15: 73556369-73556369 on Build GRCh38
rs377361229 Chr.15: 73556345-73556345 on Build GRCh38
rs74488445 Chr.15: 73556299-73556299 on Build GRCh38
rs774138589 Chr.15: 73556365-73556365 on Build GRCh38
rs942890681 Chr.15: 73556212-73556212 on Build GRCh38
rs762071708 Chr.15: 73556378-73556378 on Build GRCh38

View Full Product Details