Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 100140843-100141068 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRMT13
Entrez Gene ID
54482
Gene Name
tRNA methyltransferase 13 homolog
Gene Aliases
CCDC76
UniGene
Hs.729095
Gene Chromosome Location
Chr.1: 100133080-100150498 on Build GRCh38
Transcripts
NM_019083.2 NR_135078.1 XM_005270945.3 XM_005270946.2 XM_017001504.1 XM_017001505.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4019796 c.637A>G p.I213V Chr.1: 100140987-100140987 on Build GRCh38
1747556 c.561T>A p.V187V Chr.1: 100140911-100140911 on Build GRCh38
3965666 c.515G>A p.G172D Chr.1: 100140865-100140865 on Build GRCh38
4597251 c.569G>T p.G190V Chr.1: 100140919-100140919 on Build GRCh38
4597250 c.551G>T p.R184I Chr.1: 100140901-100140901 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750681816 Chr.1: 100140953-100140953 on Build GRCh38
rs200758373 Chr.1: 100140938-100140938 on Build GRCh38
rs781384174 Chr.1: 100140948-100140948 on Build GRCh38
rs779576942 Chr.1: 100140979-100140979 on Build GRCh38
rs758244108 Chr.1: 100140944-100140944 on Build GRCh38
rs771793798 Chr.1: 100140904-100140904 on Build GRCh38
rs34001526 Chr.1: 100140877-100140878 on Build GRCh38
rs760427282 Chr.1: 100140990-100140990 on Build GRCh38
rs767403450 Chr.1: 100140983-100140984 on Build GRCh38
rs537478308 Chr.1: 100140923-100140923 on Build GRCh38
rs375500247 Chr.1: 100140937-100140937 on Build GRCh38
rs756122850 Chr.1: 100140956-100140956 on Build GRCh38
rs201445065 Chr.1: 100140983-100140983 on Build GRCh38
rs184599018 Chr.1: 100140963-100140963 on Build GRCh38
rs762703092 Chr.1: 100141036-100141036 on Build GRCh38
rs753699072 Chr.1: 100140934-100140934 on Build GRCh38
rs146877147 Chr.1: 100140945-100140945 on Build GRCh38
rs772970276 Chr.1: 100140986-100140986 on Build GRCh38
rs762272288 Chr.1: 100140893-100140893 on Build GRCh38
rs920915278 Chr.1: 100140939-100140939 on Build GRCh38
rs1039597765 Chr.1: 100140992-100140992 on Build GRCh38
rs775469551 Chr.1: 100141019-100141019 on Build GRCh38
rs372108290 Chr.1: 100141007-100141007 on Build GRCh38
rs943798430 Chr.1: 100140922-100140922 on Build GRCh38
rs765141854 Chr.1: 100141008-100141008 on Build GRCh38
rs539584360 Chr.1: 100140980-100140980 on Build GRCh38
rs1042971703 Chr.1: 100140993-100140993 on Build GRCh38
rs80105468 Chr.1: 100140998-100140998 on Build GRCh38
rs140975434 Chr.1: 100140995-100140995 on Build GRCh38
rs148398212 Chr.1: 100140888-100140888 on Build GRCh38
rs752427125 Chr.1: 100140943-100140943 on Build GRCh38
rs983602611 Chr.1: 100140872-100140872 on Build GRCh38
rs773058210 Chr.1: 100140905-100140905 on Build GRCh38
rs144357749 Chr.1: 100140960-100140960 on Build GRCh38
rs760101160 Chr.1: 100140910-100140910 on Build GRCh38
rs769139529 Chr.1: 100140971-100140971 on Build GRCh38

View Full Product Details