Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 68197249-68197492 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALML4
Entrez Gene ID
91860
Gene Name
calmodulin like 4
Gene Aliases
NY-BR-20
UniGene
Hs.709550
Gene Chromosome Location
Chr.15: 68190705-68206110 on Build GRCh38
Transcripts
NM_001031733.2 NM_001286694.1 NM_001286695.1 NM_033429.2 NR_104583.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3678251 c.461C>A p.T154K Chr.15: 68197472-68197472 on Build GRCh38
5913647 c.490G>A p.E164K Chr.15: 68197443-68197443 on Build GRCh38
3988089 c.448C>T p.R150W Chr.15: 68197485-68197485 on Build GRCh38
1238416 c.449G>A p.R150Q Chr.15: 68197484-68197484 on Build GRCh38
1247260 c.493+6_493+7insTGTT p.? Chr.15: 68197433-68197434 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761954635 Chr.15: 68197320-68197320 on Build GRCh38
rs754686725 Chr.15: 68197445-68197445 on Build GRCh38
rs766244711 Chr.15: 68197424-68197424 on Build GRCh38
rs750425515 Chr.15: 68197413-68197413 on Build GRCh38
rs368831643 Chr.15: 68197423-68197423 on Build GRCh38
rs984564696 Chr.15: 68197325-68197325 on Build GRCh38
rs781329754 Chr.15: 68197465-68197465 on Build GRCh38
rs907713521 Chr.15: 68197326-68197326 on Build GRCh38
rs200077073 Chr.15: 68197311-68197312 on Build GRCh38
rs761007797 Chr.15: 68197401-68197401 on Build GRCh38
rs764173580 Chr.15: 68197403-68197403 on Build GRCh38
rs28507642 Chr.15: 68197410-68197410 on Build GRCh38
rs776725963 Chr.15: 68197404-68197404 on Build GRCh38
rs937388465 Chr.15: 68197360-68197360 on Build GRCh38
rs999435961 Chr.15: 68197271-68197271 on Build GRCh38
rs1023482324 Chr.15: 68197300-68197300 on Build GRCh38
rs199952882 Chr.15: 68197470-68197470 on Build GRCh38
rs923756006 Chr.15: 68197416-68197416 on Build GRCh38
rs946446434 Chr.15: 68197407-68197407 on Build GRCh38
rs780923053 Chr.15: 68197456-68197456 on Build GRCh38
rs35247364 Chr.15: 68197294-68197295 on Build GRCh38
rs772559404 Chr.15: 68197393-68197393 on Build GRCh38
rs558497506 Chr.15: 68197328-68197328 on Build GRCh38
rs755636457 Chr.15: 68197431-68197431 on Build GRCh38
rs965779148 Chr.15: 68197304-68197304 on Build GRCh38
rs566530501 Chr.15: 68197309-68197309 on Build GRCh38
rs554953385 Chr.15: 68197471-68197471 on Build GRCh38
rs2280217 Chr.15: 68197472-68197472 on Build GRCh38
rs765450917 Chr.15: 68197324-68197324 on Build GRCh38
rs775689596 Chr.15: 68197396-68197396 on Build GRCh38
rs751260030 Chr.15: 68197441-68197441 on Build GRCh38
rs370364706 Chr.15: 68197461-68197461 on Build GRCh38
rs538653218 Chr.15: 68197323-68197323 on Build GRCh38
rs895784349 Chr.15: 68197279-68197280 on Build GRCh38
rs928611100 Chr.15: 68197347-68197347 on Build GRCh38
rs1019018236 Chr.15: 68197299-68197299 on Build GRCh38
rs751432197 Chr.15: 68197327-68197327 on Build GRCh38

View Full Product Details