Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 167126789-167127062 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP27
Entrez Gene ID
92235
Gene Name
dual specificity phosphatase 27 (putative)
Gene Aliases
-
UniGene
Hs.632462
Gene Chromosome Location
Chr.1: 167094048-167129165 on Build GRCh38
Transcripts
NM_001080426.1 XM_005245603.3 XM_011510146.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4024785 c.1670A>T p.H557L Chr.1: 167126801-167126801 on Build GRCh38
4763403 c.1745C>G p.S582C Chr.1: 167126876-167126876 on Build GRCh38
4532550 c.1895G>A p.R632K Chr.1: 167127026-167127026 on Build GRCh38
1743217 c.1775G>T p.W592L Chr.1: 167126906-167126906 on Build GRCh38
4024786 c.1901G>A p.R634Q Chr.1: 167127032-167127032 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372727539 Chr.1: 167126832-167126832 on Build GRCh38
rs747943857 Chr.1: 167127029-167127029 on Build GRCh38
rs529604128 Chr.1: 167126936-167126936 on Build GRCh38
rs781679020 Chr.1: 167126847-167126847 on Build GRCh38
rs545061292 Chr.1: 167126981-167126981 on Build GRCh38
rs1002949988 Chr.1: 167126959-167126959 on Build GRCh38
rs146837441 Chr.1: 167126833-167126833 on Build GRCh38
rs768511723 Chr.1: 167126900-167126900 on Build GRCh38
rs202017285 Chr.1: 167127038-167127038 on Build GRCh38
rs749647831 Chr.1: 167126865-167126865 on Build GRCh38
rs374488804 Chr.1: 167126984-167126984 on Build GRCh38
rs778336860 Chr.1: 167126970-167126970 on Build GRCh38
rs750042465 Chr.1: 167126918-167126918 on Build GRCh38
rs757387713 Chr.1: 167126872-167126872 on Build GRCh38
rs562950788 Chr.1: 167126920-167126920 on Build GRCh38
rs867643169 Chr.1: 167126974-167126974 on Build GRCh38
rs771122589 Chr.1: 167126877-167126877 on Build GRCh38
rs149689505 Chr.1: 167126931-167126931 on Build GRCh38
rs776349275 Chr.1: 167126901-167126901 on Build GRCh38
rs368596520 Chr.1: 167126828-167126828 on Build GRCh38
rs757616235 Chr.1: 167126982-167126982 on Build GRCh38
rs906677033 Chr.1: 167126906-167126906 on Build GRCh38
rs549179166 Chr.1: 167127032-167127032 on Build GRCh38
rs753215878 Chr.1: 167126942-167126942 on Build GRCh38
rs976666073 Chr.1: 167127014-167127014 on Build GRCh38
rs1011449034 Chr.1: 167126971-167126971 on Build GRCh38
rs747073864 Chr.1: 167127005-167127005 on Build GRCh38
rs772284726 Chr.1: 167126993-167126993 on Build GRCh38
rs1037207381 Chr.1: 167126874-167126874 on Build GRCh38
rs772856160 Chr.1: 167127031-167127031 on Build GRCh38
rs772233606 Chr.1: 167126891-167126891 on Build GRCh38
rs761664135 Chr.1: 167126907-167126907 on Build GRCh38
rs756472833 Chr.1: 167126863-167126863 on Build GRCh38
rs760626149 Chr.1: 167126899-167126899 on Build GRCh38
rs753162739 Chr.1: 167126858-167126858 on Build GRCh38
rs775602117 Chr.1: 167126894-167126894 on Build GRCh38
rs144793075 Chr.1: 167126889-167126889 on Build GRCh38
rs746051072 Chr.1: 167126992-167126992 on Build GRCh38
rs779159793 Chr.1: 167126878-167126878 on Build GRCh38
rs749967832 Chr.1: 167126835-167126835 on Build GRCh38
rs753536435 Chr.1: 167126820-167126820 on Build GRCh38
rs756668951 Chr.1: 167126956-167126956 on Build GRCh38
rs768544982 Chr.1: 167127020-167127020 on Build GRCh38
rs188895908 Chr.1: 167127004-167127004 on Build GRCh38
rs764553686 Chr.1: 167127039-167127039 on Build GRCh38
rs776600431 Chr.1: 167127022-167127022 on Build GRCh38
rs956126023 Chr.1: 167126985-167126985 on Build GRCh38
rs987900395 Chr.1: 167127028-167127028 on Build GRCh38
rs369939829 Chr.1: 167126881-167126881 on Build GRCh38
rs573913763 Chr.1: 167126819-167126819 on Build GRCh38
rs769590965 Chr.1: 167127030-167127030 on Build GRCh38
rs896852713 Chr.1: 167126861-167126861 on Build GRCh38
rs757874206 Chr.1: 167126842-167126842 on Build GRCh38
rs764839180 Chr.1: 167126913-167126913 on Build GRCh38

View Full Product Details