Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 32551327-32551582 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AVL9
Entrez Gene ID
23080
Gene Name
AVL9 cell migration associated
Gene Aliases
KIAA0241
UniGene
Hs.128056
Gene Chromosome Location
Chr.7: 32495426-32588741 on Build GRCh38
Transcripts
NM_015060.2 XM_005249668.2 XM_005249669.2 XM_005249670.2 XM_005249671.4 XM_011515227.2 XM_017011891.1 XM_017011892.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3228123 c.462G>T p.K154N Chr.7: 32551423-32551423 on Build GRCh38
4940755 c.416A>G p.H139R Chr.7: 32551377-32551377 on Build GRCh38
3228122 c.462G>T p.K154N Chr.7: 32551423-32551423 on Build GRCh38
3394717 c.462+8T>C p.? Chr.7: 32551431-32551431 on Build GRCh38
5467125 c.452C>A p.S151Y Chr.7: 32551413-32551413 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139597099 Chr.7: 32551357-32551357 on Build GRCh38
rs764039028 Chr.7: 32551432-32551432 on Build GRCh38
rs751057561 Chr.7: 32551435-32551435 on Build GRCh38
rs765112789 Chr.7: 32551369-32551369 on Build GRCh38
rs371602120 Chr.7: 32551463-32551463 on Build GRCh38
rs140931579 Chr.7: 32551532-32551532 on Build GRCh38
rs878973321 Chr.7: 32551488-32551488 on Build GRCh38
rs3213973 Chr.7: 32551546-32551546 on Build GRCh38
rs905373246 Chr.7: 32551377-32551377 on Build GRCh38
rs772478670 Chr.7: 32551460-32551460 on Build GRCh38
rs377011753 Chr.7: 32551378-32551378 on Build GRCh38
rs748484889 Chr.7: 32551456-32551456 on Build GRCh38
rs1040570215 Chr.7: 32551529-32551529 on Build GRCh38
rs957027996 Chr.7: 32551450-32551450 on Build GRCh38
rs759454167 Chr.7: 32551366-32551366 on Build GRCh38
rs559245652 Chr.7: 32551522-32551522 on Build GRCh38
rs779148409 Chr.7: 32551449-32551449 on Build GRCh38
rs758275867 Chr.7: 32551404-32551404 on Build GRCh38
rs1002388854 Chr.7: 32551497-32551497 on Build GRCh38
rs924237005 Chr.7: 32551436-32551436 on Build GRCh38
rs3213972 Chr.7: 32551523-32551523 on Build GRCh38
rs1035249915 Chr.7: 32551506-32551506 on Build GRCh38
rs12672899 Chr.7: 32551533-32551533 on Build GRCh38
rs780867418 Chr.7: 32551446-32551446 on Build GRCh38
rs755399546 Chr.7: 32551448-32551448 on Build GRCh38
rs1025814132 Chr.7: 32551537-32551537 on Build GRCh38

View Full Product Details