Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 60149278-60149511 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHRS7
Entrez Gene ID
51635
Gene Name
dehydrogenase/reductase 7
Gene Aliases
CGI-86, SDR34C1, retDSR4, retSDR4
UniGene
Hs.59719
Gene Chromosome Location
Chr.14: 60144119-60169864 on Build GRCh38
Transcripts
NM_001322280.1 NM_001322281.1 NM_001322282.1 NM_016029.3 XM_017021368.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3815016 c.873C>T p.F291F Chr.14: 60149452-60149452 on Build GRCh38
1370335 c.942G>T p.K314N Chr.14: 60149383-60149383 on Build GRCh38
389135 c.860C>T p.S287L Chr.14: 60149465-60149465 on Build GRCh38
433193 c.825C>T p.I275I Chr.14: 60149500-60149500 on Build GRCh38
1238668 c.956A>G p.N319S Chr.14: 60149369-60149369 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766709468 Chr.14: 60149404-60149404 on Build GRCh38
rs753484563 Chr.14: 60149395-60149395 on Build GRCh38
rs909067850 Chr.14: 60149430-60149430 on Build GRCh38
rs774883587 Chr.14: 60149377-60149377 on Build GRCh38
rs966403262 Chr.14: 60149362-60149362 on Build GRCh38
rs777910825 Chr.14: 60149470-60149470 on Build GRCh38
rs749157801 Chr.14: 60149359-60149359 on Build GRCh38
rs1048906207 Chr.14: 60149411-60149411 on Build GRCh38
rs112959466 Chr.14: 60149351-60149351 on Build GRCh38
rs143781219 Chr.14: 60149357-60149357 on Build GRCh38
rs763659152 Chr.14: 60149388-60149388 on Build GRCh38
rs759513450 Chr.14: 60149372-60149372 on Build GRCh38
rs755513672 Chr.14: 60149393-60149395 on Build GRCh38
rs771915686 Chr.14: 60149478-60149478 on Build GRCh38
rs752736783 Chr.14: 60149456-60149456 on Build GRCh38
rs781690782 Chr.14: 60149449-60149449 on Build GRCh38
rs760173791 Chr.14: 60149378-60149378 on Build GRCh38
rs758822114 Chr.14: 60149403-60149403 on Build GRCh38
rs778880826 Chr.14: 60149474-60149474 on Build GRCh38
rs111725318 Chr.14: 60149479-60149479 on Build GRCh38
rs757644383 Chr.14: 60149360-60149360 on Build GRCh38
rs765793400 Chr.14: 60149398-60149398 on Build GRCh38
rs752102316 Chr.14: 60149406-60149406 on Build GRCh38
rs1016271719 Chr.14: 60149384-60149384 on Build GRCh38
rs530458404 Chr.14: 60149347-60149347 on Build GRCh38
rs976889155 Chr.14: 60149414-60149414 on Build GRCh38
rs372530611 Chr.14: 60149312-60149312 on Build GRCh38
rs747502274 Chr.14: 60149343-60149343 on Build GRCh38
rs748221259 Chr.14: 60149349-60149349 on Build GRCh38
rs922812738 Chr.14: 60149421-60149421 on Build GRCh38
rs755472688 Chr.14: 60149422-60149422 on Build GRCh38
rs411880 Chr.14: 60149324-60149324 on Build GRCh38
rs755751780 Chr.14: 60149316-60149316 on Build GRCh38
rs376507004 Chr.14: 60149458-60149458 on Build GRCh38
rs770886627 Chr.14: 60149472-60149472 on Build GRCh38
rs199694801 Chr.14: 60149327-60149327 on Build GRCh38
rs147897164 Chr.14: 60149325-60149325 on Build GRCh38
rs758655714 Chr.14: 60149364-60149365 on Build GRCh38
rs940548010 Chr.14: 60149431-60149431 on Build GRCh38
rs138163363 Chr.14: 60149352-60149352 on Build GRCh38
rs113852712 Chr.14: 60149350-60149350 on Build GRCh38
rs932394659 Chr.14: 60149435-60149435 on Build GRCh38
rs770833155 Chr.14: 60149365-60149365 on Build GRCh38
rs371048157 Chr.14: 60149375-60149375 on Build GRCh38
rs774475443 Chr.14: 60149371-60149371 on Build GRCh38
rs1036663489 Chr.14: 60149436-60149436 on Build GRCh38
rs745607417 Chr.14: 60149475-60149475 on Build GRCh38
rs767005889 Chr.14: 60149354-60149354 on Build GRCh38
rs763752104 Chr.14: 60149321-60149321 on Build GRCh38

View Full Product Details