Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 120660868-120661109 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CABP1
Entrez Gene ID
9478
Gene Name
calcium binding protein 1
Gene Aliases
CALBRAIN, HCALB_BR
UniGene
Hs.458482
Gene Chromosome Location
Chr.12: 120640552-120680869 on Build GRCh38
Transcripts
NM_001033677.1 NM_004276.4 NM_031205.3 XM_017020235.1 XM_017020236.1 XM_017020237.1 XM_017020238.1 XM_017020239.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5932977 c.952G>A p.G318S Chr.12: 120661083-120661083 on Build GRCh38
5932978 c.523G>A p.G175S Chr.12: 120661083-120661083 on Build GRCh38
458250 c.946A>G p.T316A Chr.12: 120661077-120661077 on Build GRCh38
1247060 c.953G>A p.G318D Chr.12: 120661084-120661084 on Build GRCh38
6375542 c.952G>T p.G318C Chr.12: 120661083-120661083 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1031417418 Chr.12: 120660940-120660940 on Build GRCh38
rs1034670505 Chr.12: 120660964-120660964 on Build GRCh38
rs144867541 Chr.12: 120660978-120660978 on Build GRCh38
rs371724756 Chr.12: 120661029-120661029 on Build GRCh38
rs375745402 Chr.12: 120661063-120661063 on Build GRCh38
rs1022596307 Chr.12: 120660900-120660900 on Build GRCh38
rs951923165 Chr.12: 120660998-120660998 on Build GRCh38
rs763964777 Chr.12: 120661027-120661027 on Build GRCh38
rs201080747 Chr.12: 120661042-120661042 on Build GRCh38
rs571942596 Chr.12: 120660906-120660906 on Build GRCh38
rs961674853 Chr.12: 120660975-120660975 on Build GRCh38
rs924639042 Chr.12: 120661068-120661068 on Build GRCh38
rs547631579 Chr.12: 120660921-120660921 on Build GRCh38
rs746113332 Chr.12: 120661082-120661082 on Build GRCh38
rs753638328 Chr.12: 120661028-120661028 on Build GRCh38
rs904909420 Chr.12: 120660917-120660917 on Build GRCh38
rs371380920 Chr.12: 120661061-120661061 on Build GRCh38
rs746503423 Chr.12: 120661041-120661041 on Build GRCh38
rs756542168 Chr.12: 120661037-120661037 on Build GRCh38
rs778357660 Chr.12: 120661056-120661056 on Build GRCh38
rs976153016 Chr.12: 120661067-120661067 on Build GRCh38
rs756659075 Chr.12: 120660973-120660974 on Build GRCh38
rs1013461959 Chr.12: 120660893-120660893 on Build GRCh38
rs893347044 Chr.12: 120660892-120660892 on Build GRCh38
rs775605173 Chr.12: 120661021-120661021 on Build GRCh38
rs749698253 Chr.12: 120661058-120661058 on Build GRCh38
rs188304140 Chr.12: 120661035-120661035 on Build GRCh38
rs978545755 Chr.12: 120660933-120660933 on Build GRCh38
rs750094045 Chr.12: 120661033-120661033 on Build GRCh38
rs372659944 Chr.12: 120661049-120661049 on Build GRCh38
rs971995598 Chr.12: 120660976-120660976 on Build GRCh38
rs760666044 Chr.12: 120661026-120661026 on Build GRCh38
rs534409658 Chr.12: 120660907-120660907 on Build GRCh38
rs764966319 Chr.12: 120661031-120661031 on Build GRCh38
rs568427160 Chr.12: 120660914-120660914 on Build GRCh38
rs1003227070 Chr.12: 120660944-120660944 on Build GRCh38
rs552129817 Chr.12: 120660894-120660894 on Build GRCh38

View Full Product Details