Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 75675516-75675747 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSPG4
Entrez Gene ID
1464
Gene Name
chondroitin sulfate proteoglycan 4
Gene Aliases
HMW-MAA, MCSP, MCSPG, MEL-CSPG, MSK16, NG2
UniGene
Hs.513044
Gene Chromosome Location
Chr.15: 75674322-75712848 on Build GRCh38
Transcripts
NM_001897.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6078015 c.6858C>A p.L2286L Chr.15: 75675661-75675661 on Build GRCh38
1608697 c.6827G>A p.R2276H Chr.15: 75675692-75675692 on Build GRCh38
4556103 c.6799G>T p.G2267C Chr.15: 75675720-75675720 on Build GRCh38
4057003 c.6793C>T p.R2265C Chr.15: 75675726-75675726 on Build GRCh38
1202511 c.6863C>G p.A2288G Chr.15: 75675656-75675656 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774145739 Chr.15: 75675726-75675726 on Build GRCh38
rs781171777 Chr.15: 75675556-75675556 on Build GRCh38
rs769627899 Chr.15: 75675562-75675562 on Build GRCh38
rs779833639 Chr.15: 75675682-75675682 on Build GRCh38
rs530682211 Chr.15: 75675638-75675638 on Build GRCh38
rs115161109 Chr.15: 75675583-75675583 on Build GRCh38
rs144719140 Chr.15: 75675652-75675652 on Build GRCh38
rs768486425 Chr.15: 75675687-75675687 on Build GRCh38
rs934553944 Chr.15: 75675651-75675651 on Build GRCh38
rs773175028 Chr.15: 75675703-75675703 on Build GRCh38
rs762651688 Chr.15: 75675648-75675648 on Build GRCh38
rs749456642 Chr.15: 75675578-75675578 on Build GRCh38
rs3203767 Chr.15: 75675713-75675713 on Build GRCh38
rs559710810 Chr.15: 75675665-75675665 on Build GRCh38
rs748610777 Chr.15: 75675571-75675571 on Build GRCh38
rs771369858 Chr.15: 75675714-75675714 on Build GRCh38
rs757214708 Chr.15: 75675537-75675537 on Build GRCh38
rs749101671 Chr.15: 75675684-75675684 on Build GRCh38
rs761175264 Chr.15: 75675709-75675709 on Build GRCh38
rs779770942 Chr.15: 75675564-75675564 on Build GRCh38
rs779075757 Chr.15: 75675692-75675692 on Build GRCh38
rs186191601 Chr.15: 75675593-75675593 on Build GRCh38
rs765597004 Chr.15: 75675725-75675725 on Build GRCh38
rs756800409 Chr.15: 75675655-75675655 on Build GRCh38
rs780637386 Chr.15: 75675664-75675664 on Build GRCh38
rs772178990 Chr.15: 75675588-75675588 on Build GRCh38
rs772175577 Chr.15: 75675702-75675702 on Build GRCh38
rs563285050 Chr.15: 75675594-75675594 on Build GRCh38
rs969695409 Chr.15: 75675577-75675577 on Build GRCh38
rs545688154 Chr.15: 75675720-75675720 on Build GRCh38
rs148118120 Chr.15: 75675721-75675721 on Build GRCh38
rs745837181 Chr.15: 75675559-75675559 on Build GRCh38
rs143005137 Chr.15: 75675584-75675584 on Build GRCh38
rs748434385 Chr.15: 75675693-75675693 on Build GRCh38
rs756019878 Chr.15: 75675673-75675673 on Build GRCh38

View Full Product Details