Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 183446828-183447083 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDKN2AIP
Entrez Gene ID
55602
Gene Name
CDKN2A interacting protein
Gene Aliases
CARF
UniGene
Hs.592508
Gene Chromosome Location
Chr.4: 183444591-183449064 on Build GRCh38
Transcripts
NM_001317343.1 NM_017632.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6882951 c.1257G>A p.E419E Chr.4: 183446941-183446941 on Build GRCh38
3602379 c.1190C>T p.A397V Chr.4: 183446874-183446874 on Build GRCh38
5742606 c.1314_1315insT p.N441fs*2 Chr.4: 183446998-183446999 on Build GRCh38
6625729 c.1314delT p.F440fs*13 Chr.4: 183446998-183446998 on Build GRCh38
5455287 c.1274C>A p.S425Y Chr.4: 183446958-183446958 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138463372 Chr.4: 183446956-183446956 on Build GRCh38
rs771344065 Chr.4: 183446882-183446884 on Build GRCh38
rs941238397 Chr.4: 183446880-183446880 on Build GRCh38
rs753862628 Chr.4: 183446919-183446919 on Build GRCh38
rs760469515 Chr.4: 183446913-183446913 on Build GRCh38
rs141991851 Chr.4: 183446920-183446920 on Build GRCh38
rs767688935 Chr.4: 183447028-183447028 on Build GRCh38
rs921136302 Chr.4: 183446904-183446904 on Build GRCh38
rs775191114 Chr.4: 183446964-183446964 on Build GRCh38
rs199963421 Chr.4: 183446995-183446995 on Build GRCh38
rs775066417 Chr.4: 183446908-183446908 on Build GRCh38
rs781564950 Chr.4: 183446933-183446933 on Build GRCh38
rs765179780 Chr.4: 183446924-183446924 on Build GRCh38
rs369366498 Chr.4: 183447043-183447043 on Build GRCh38
rs750315909 Chr.4: 183447013-183447013 on Build GRCh38
rs765276188 Chr.4: 183447005-183447005 on Build GRCh38
rs1039633713 Chr.4: 183446897-183446897 on Build GRCh38
rs373518027 Chr.4: 183447055-183447055 on Build GRCh38
rs778018161 Chr.4: 183447056-183447056 on Build GRCh38
rs150114244 Chr.4: 183446948-183446948 on Build GRCh38
rs776319496 Chr.4: 183446974-183446974 on Build GRCh38
rs779344281 Chr.4: 183446955-183446955 on Build GRCh38
rs745397785 Chr.4: 183446961-183446961 on Build GRCh38
rs763782262 Chr.4: 183446917-183446917 on Build GRCh38
rs903244741 Chr.4: 183446952-183446952 on Build GRCh38
rs759008324 Chr.4: 183446895-183446895 on Build GRCh38
rs777982179 Chr.4: 183446942-183446942 on Build GRCh38
rs749748152 Chr.4: 183446859-183446859 on Build GRCh38
rs780182422 Chr.4: 183446896-183446896 on Build GRCh38
rs768536227 Chr.4: 183446973-183446973 on Build GRCh38
rs759751969 Chr.4: 183447022-183447022 on Build GRCh38
rs200444062 Chr.4: 183446941-183446941 on Build GRCh38
rs770528042 Chr.4: 183446888-183446888 on Build GRCh38
rs755048889 Chr.4: 183446931-183446931 on Build GRCh38
rs751708633 Chr.4: 183446928-183446928 on Build GRCh38
rs773822857 Chr.4: 183446892-183446892 on Build GRCh38

View Full Product Details