Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 130207888-130208157 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SSMEM1
Entrez Gene ID
136263
Gene Name
serine rich single-pass membrane protein 1
Gene Aliases
C7orf45
UniGene
Hs.351816
Gene Chromosome Location
Chr.7: 130205399-130216869 on Build GRCh38
Transcripts
NM_145268.3 XM_005250141.3 XM_011515795.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4934383 c.25T>G p.W9G Chr.7: 130207935-130207935 on Build GRCh38
4931589 c.183G>T p.W61C Chr.7: 130208093-130208093 on Build GRCh38
6866422 c.126G>A p.L42L Chr.7: 130208036-130208036 on Build GRCh38
1448041 c.30delG p.V11fs*1 Chr.7: 130207940-130207940 on Build GRCh38
5919237 c.17C>T p.S6F Chr.7: 130207927-130207927 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763806644 Chr.7: 130207970-130207970 on Build GRCh38
rs770755410 Chr.7: 130208034-130208034 on Build GRCh38
rs749707311 Chr.7: 130207920-130207920 on Build GRCh38
rs764983904 Chr.7: 130208055-130208055 on Build GRCh38
rs779152211 Chr.7: 130208020-130208020 on Build GRCh38
rs780147118 Chr.7: 130208005-130208005 on Build GRCh38
rs750500555 Chr.7: 130207917-130207917 on Build GRCh38
rs371191805 Chr.7: 130207950-130207950 on Build GRCh38
rs868126292 Chr.7: 130207927-130207927 on Build GRCh38
rs779288452 Chr.7: 130207924-130207924 on Build GRCh38
rs977079440 Chr.7: 130208023-130208023 on Build GRCh38
rs201225197 Chr.7: 130207928-130207928 on Build GRCh38
rs148378256 Chr.7: 130207956-130207956 on Build GRCh38
rs756159838 Chr.7: 130207997-130207997 on Build GRCh38
rs775003462 Chr.7: 130208068-130208068 on Build GRCh38
rs377008348 Chr.7: 130208015-130208015 on Build GRCh38
rs140531337 Chr.7: 130207963-130207963 on Build GRCh38
rs746675089 Chr.7: 130207933-130207933 on Build GRCh38
rs778451243 Chr.7: 130207947-130207947 on Build GRCh38
rs186454728 Chr.7: 130207951-130207951 on Build GRCh38
rs921391864 Chr.7: 130208111-130208111 on Build GRCh38
rs111931380 Chr.7: 130207939-130207939 on Build GRCh38
rs749182968 Chr.7: 130208075-130208075 on Build GRCh38
rs753411853 Chr.7: 130207955-130207955 on Build GRCh38
rs750553921 Chr.7: 130207994-130207994 on Build GRCh38
rs759075254 Chr.7: 130208054-130208054 on Build GRCh38
rs756675106 Chr.7: 130207919-130207919 on Build GRCh38
rs534971170 Chr.7: 130208112-130208112 on Build GRCh38
rs774192176 Chr.7: 130208097-130208097 on Build GRCh38
rs766775566 Chr.7: 130207953-130207953 on Build GRCh38
rs762087081 Chr.7: 130207990-130207990 on Build GRCh38
rs759093657 Chr.7: 130207946-130207946 on Build GRCh38
rs770643207 Chr.7: 130207936-130207936 on Build GRCh38
rs748520984 Chr.7: 130208021-130208021 on Build GRCh38
rs372434401 Chr.7: 130207992-130207992 on Build GRCh38
rs776336519 Chr.7: 130207937-130207937 on Build GRCh38
rs968341925 Chr.7: 130208007-130208007 on Build GRCh38
rs768630597 Chr.7: 130208094-130208094 on Build GRCh38
rs751298433 Chr.7: 130207980-130207980 on Build GRCh38

View Full Product Details