Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 69133403-69133674 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UTP4
Entrez Gene ID
84916
Gene Name
UTP4, small subunit processome component
Gene Aliases
CIRH1A, CIRHIN, NAIC, TEX292
UniGene
Hs.461113
Gene Chromosome Location
Chr.16: 69132596-69169034 on Build GRCh38
Transcripts
NM_001318391.1 NM_032830.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5043186 c.4G>T p.G2C Chr.16: 69133463-69133463 on Build GRCh38
6021469 c.89G>C p.R30T Chr.16: 69133548-69133548 on Build GRCh38
7179454 c.115A>G p.T39A Chr.16: 69133574-69133574 on Build GRCh38
6250319 c.66G>T p.V22V Chr.16: 69133525-69133525 on Build GRCh38
7001563 c.29_30insAGTACG p.R10_F11insVR Chr.16: 69133488-69133489 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764719731 Chr.16: 69133483-69133496 on Build GRCh38
rs141243868 Chr.16: 69133516-69133516 on Build GRCh38
rs767459090 Chr.16: 69133510-69133510 on Build GRCh38
rs995446622 Chr.16: 69133526-69133526 on Build GRCh38
rs763383121 Chr.16: 69133577-69133577 on Build GRCh38
rs770495589 Chr.16: 69133550-69133550 on Build GRCh38
rs895142389 Chr.16: 69133495-69133495 on Build GRCh38
rs754816537 Chr.16: 69133519-69133519 on Build GRCh38
rs561863834 Chr.16: 69133508-69133508 on Build GRCh38
rs778685504 Chr.16: 69133520-69133520 on Build GRCh38
rs35755396 Chr.16: 69133562-69133563 on Build GRCh38
rs777224876 Chr.16: 69133443-69133443 on Build GRCh38
rs771380331 Chr.16: 69133439-69133439 on Build GRCh38
rs994446961 Chr.16: 69133552-69133552 on Build GRCh38
rs1030203914 Chr.16: 69133574-69133574 on Build GRCh38
rs751741576 Chr.16: 69133470-69133470 on Build GRCh38
rs373005279 Chr.16: 69133466-69133466 on Build GRCh38
rs775944744 Chr.16: 69133551-69133551 on Build GRCh38
rs755010673 Chr.16: 69133641-69133641 on Build GRCh38
rs941383539 Chr.16: 69133518-69133518 on Build GRCh38
rs747933459 Chr.16: 69133527-69133527 on Build GRCh38
rs9929653 Chr.16: 69133642-69133642 on Build GRCh38
rs774749174 Chr.16: 69133603-69133603 on Build GRCh38
rs368784646 Chr.16: 69133448-69133448 on Build GRCh38
rs776051732 Chr.16: 69133565-69133565 on Build GRCh38
rs776980441 Chr.16: 69133627-69133627 on Build GRCh38
rs770416587 Chr.16: 69133562-69133562 on Build GRCh38
rs765645929 Chr.16: 69133453-69133453 on Build GRCh38
rs146966335 Chr.16: 69133533-69133533 on Build GRCh38
rs761895192 Chr.16: 69133629-69133629 on Build GRCh38
rs765771251 Chr.16: 69133476-69133476 on Build GRCh38
rs760665575 Chr.16: 69133439-69133440 on Build GRCh38
rs750404559 Chr.16: 69133632-69133632 on Build GRCh38
rs1004311751 Chr.16: 69133460-69133460 on Build GRCh38
rs775927023 Chr.16: 69133454-69133454 on Build GRCh38
rs74886292 Chr.16: 69133557-69133557 on Build GRCh38
rs767650795 Chr.16: 69133630-69133630 on Build GRCh38
rs764454732 Chr.16: 69133593-69133593 on Build GRCh38
rs1006586645 Chr.16: 69133628-69133628 on Build GRCh38
rs757331854 Chr.16: 69133477-69133477 on Build GRCh38
rs758762300 Chr.16: 69133572-69133572 on Build GRCh38
rs753272736 Chr.16: 69133542-69133542 on Build GRCh38
rs746601254 Chr.16: 69133546-69133546 on Build GRCh38

View Full Product Details