Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 31626996-31627221 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B4GALT6
Entrez Gene ID
9331
Gene Name
beta-1,4-galactosyltransferase 6
Gene Aliases
B4Gal-T6, beta4Gal-T6
UniGene
Hs.591063
Gene Chromosome Location
Chr.18: 31622246-31724670 on Build GRCh38
Transcripts
NM_004775.3 XM_005258387.4 XM_006722578.3 XM_006722579.3 XM_017026088.1 XM_017026089.1 XM_017026090.1 XM_017026091.1 XM_017026092.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
279106 c.839T>G p.I280S Chr.18: 31627059-31627059 on Build GRCh38
708079 c.866G>A p.G289E Chr.18: 31627032-31627032 on Build GRCh38
987648 c.848T>A p.F283Y Chr.18: 31627050-31627050 on Build GRCh38
3422121 c.795T>G p.F265L Chr.18: 31627103-31627103 on Build GRCh38
3388418 c.857C>G p.A286G Chr.18: 31627041-31627041 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs906715504 Chr.18: 31627140-31627140 on Build GRCh38
rs549667383 Chr.18: 31627043-31627043 on Build GRCh38
rs535446741 Chr.18: 31627176-31627176 on Build GRCh38
rs774347913 Chr.18: 31627121-31627121 on Build GRCh38
rs141729397 Chr.18: 31627194-31627194 on Build GRCh38
rs767362690 Chr.18: 31627129-31627129 on Build GRCh38
rs1026123 Chr.18: 31627192-31627192 on Build GRCh38
rs747287363 Chr.18: 31627056-31627056 on Build GRCh38
rs765484260 Chr.18: 31627126-31627126 on Build GRCh38
rs1057394568 Chr.18: 31627091-31627091 on Build GRCh38
rs201572227 Chr.18: 31627126-31627126 on Build GRCh38
rs760527944 Chr.18: 31627162-31627162 on Build GRCh38
rs776811806 Chr.18: 31627088-31627088 on Build GRCh38
rs766195137 Chr.18: 31627173-31627173 on Build GRCh38
rs771183168 Chr.18: 31627058-31627058 on Build GRCh38
rs772924057 Chr.18: 31627136-31627136 on Build GRCh38
rs763263372 Chr.18: 31627156-31627158 on Build GRCh38
rs746005124 Chr.18: 31627106-31627106 on Build GRCh38
rs184712194 Chr.18: 31627184-31627184 on Build GRCh38
rs748538956 Chr.18: 31627031-31627031 on Build GRCh38
rs752966754 Chr.18: 31627133-31627136 on Build GRCh38
rs769858727 Chr.18: 31627111-31627111 on Build GRCh38
rs959481962 Chr.18: 31627188-31627188 on Build GRCh38
rs1004150146 Chr.18: 31627179-31627179 on Build GRCh38

View Full Product Details