Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 210216380-210216613 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACADL
Entrez Gene ID
33
Gene Name
acyl-CoA dehydrogenase, long chain
Gene Aliases
ACAD4, LCAD
UniGene
Hs.471277
Gene Chromosome Location
Chr.2: 210187990-210225491 on Build GRCh38
Transcripts
NM_001608.3 XM_005246517.4 XM_017003955.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4090764 c.399T>G p.G133G Chr.2: 210216484-210216484 on Build GRCh38
1015843 c.375T>G p.A125A Chr.2: 210216508-210216508 on Build GRCh38
6591670 c.431A>G p.Y144C Chr.2: 210216452-210216452 on Build GRCh38
1405053 c.487G>A p.A163T Chr.2: 210216396-210216396 on Build GRCh38
719476 c.476C>A p.P159H Chr.2: 210216407-210216407 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766063127 Chr.2: 210216557-210216557 on Build GRCh38
rs1005993053 Chr.2: 210216464-210216464 on Build GRCh38
rs982987305 Chr.2: 210216579-210216579 on Build GRCh38
rs776182856 Chr.2: 210216558-210216558 on Build GRCh38
rs758453402 Chr.2: 210216457-210216457 on Build GRCh38
rs749663218 Chr.2: 210216493-210216493 on Build GRCh38
rs771018271 Chr.2: 210216473-210216474 on Build GRCh38
rs777872949 Chr.2: 210216466-210216466 on Build GRCh38
rs768797553 Chr.2: 210216580-210216580 on Build GRCh38
rs1007268355 Chr.2: 210216574-210216574 on Build GRCh38
rs551352082 Chr.2: 210216463-210216463 on Build GRCh38
rs753457514 Chr.2: 210216513-210216513 on Build GRCh38
rs951436317 Chr.2: 210216575-210216575 on Build GRCh38
rs774294893 Chr.2: 210216525-210216525 on Build GRCh38
rs772917241 Chr.2: 210216555-210216555 on Build GRCh38
rs754911896 Chr.2: 210216462-210216463 on Build GRCh38
rs759899944 Chr.2: 210216560-210216560 on Build GRCh38
rs760276506 Chr.2: 210216556-210216556 on Build GRCh38
rs142571448 Chr.2: 210216451-210216451 on Build GRCh38
rs748083276 Chr.2: 210216534-210216534 on Build GRCh38
rs753170102 Chr.2: 210216453-210216453 on Build GRCh38
rs184472890 Chr.2: 210216449-210216449 on Build GRCh38
rs749040869 Chr.2: 210216508-210216508 on Build GRCh38
rs753219608 Chr.2: 210216563-210216563 on Build GRCh38
rs757530795 Chr.2: 210216494-210216494 on Build GRCh38
rs372150454 Chr.2: 210216532-210216532 on Build GRCh38
rs545058005 Chr.2: 210216518-210216519 on Build GRCh38
rs571156599 Chr.2: 210216509-210216509 on Build GRCh38
rs375211389 Chr.2: 210216562-210216562 on Build GRCh38
rs777770781 Chr.2: 210216469-210216470 on Build GRCh38
rs1000484376 Chr.2: 210216515-210216515 on Build GRCh38
rs749406290 Chr.2: 210216470-210216471 on Build GRCh38
rs781402332 Chr.2: 210216501-210216501 on Build GRCh38
rs774601024 Chr.2: 210216497-210216497 on Build GRCh38
rs878858614 Chr.2: 210216564-210216564 on Build GRCh38
rs754174181 Chr.2: 210216439-210216439 on Build GRCh38
rs760472931 Chr.2: 210216418-210216418 on Build GRCh38
rs146176190 Chr.2: 210216420-210216420 on Build GRCh38
rs771675495 Chr.2: 210216551-210216551 on Build GRCh38
rs997652136 Chr.2: 210216550-210216550 on Build GRCh38
rs772561119 Chr.2: 210216520-210216521 on Build GRCh38

View Full Product Details