Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 960027-960300 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARID3A
Entrez Gene ID
1820
Gene Name
AT-rich interaction domain 3A
Gene Aliases
BRIGHT, DRIL1, DRIL3, E2FBP1
UniGene
Hs.501296
Gene Chromosome Location
Chr.19: 925733-975934 on Build GRCh38
Transcripts
NM_005224.2 XM_005259513.4 XM_005259514.4 XM_017026445.1 XM_017026446.1 XM_017026447.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1004313 c.705C>T p.L235L Chr.19: 960103-960103 on Build GRCh38
1172696 c.706G>A p.D236N Chr.19: 960104-960104 on Build GRCh38
1004314 c.705C>A p.L235L Chr.19: 960103-960103 on Build GRCh38
4890737 c.722G>T p.R241M Chr.19: 960120-960120 on Build GRCh38
6604645 c.749G>A p.S250N Chr.19: 960147-960147 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs185287816 Chr.19: 960224-960224 on Build GRCh38
rs192910888 Chr.19: 960051-960051 on Build GRCh38
rs371357347 Chr.19: 960264-960264 on Build GRCh38
rs769342047 Chr.19: 960134-960134 on Build GRCh38
rs752740985 Chr.19: 960107-960107 on Build GRCh38
rs754216623 Chr.19: 960193-960193 on Build GRCh38
rs781197545 Chr.19: 960130-960130 on Build GRCh38
rs745587090 Chr.19: 960133-960133 on Build GRCh38
rs1052867261 Chr.19: 960263-960263 on Build GRCh38
rs371658844 Chr.19: 960115-960115 on Build GRCh38
rs765407947 Chr.19: 960197-960197 on Build GRCh38
rs759317504 Chr.19: 960055-960055 on Build GRCh38
rs1637993 Chr.19: 960187-960187 on Build GRCh38
rs368236526 Chr.19: 960097-960097 on Build GRCh38
rs910458539 Chr.19: 960214-960214 on Build GRCh38
rs775171459 Chr.19: 960183-960183 on Build GRCh38
rs769478791 Chr.19: 960056-960056 on Build GRCh38
rs554289878 Chr.19: 960227-960227 on Build GRCh38
rs767944314 Chr.19: 960074-960074 on Build GRCh38
rs753208143 Chr.19: 960062-960065 on Build GRCh38
rs374854563 Chr.19: 960195-960195 on Build GRCh38
rs761152181 Chr.19: 960098-960098 on Build GRCh38
rs756663869 Chr.19: 960065-960066 on Build GRCh38
rs535131049 Chr.19: 960251-960251 on Build GRCh38
rs147646838 Chr.19: 960251-960252 on Build GRCh38
rs201781616 Chr.19: 960070-960070 on Build GRCh38
rs762479181 Chr.19: 960064-960064 on Build GRCh38
rs774863171 Chr.19: 960063-960063 on Build GRCh38
rs1042889825 Chr.19: 960240-960240 on Build GRCh38
rs2302111 Chr.19: 960277-960277 on Build GRCh38
rs746990265 Chr.19: 960121-960121 on Build GRCh38
rs567621941 Chr.19: 960232-960232 on Build GRCh38
rs904326674 Chr.19: 960250-960250 on Build GRCh38
rs369232179 Chr.19: 960202-960202 on Build GRCh38
rs1683584 Chr.19: 960266-960266 on Build GRCh38
rs770976327 Chr.19: 960127-960127 on Build GRCh38
rs779139236 Chr.19: 960104-960104 on Build GRCh38
rs982497580 Chr.19: 960237-960237 on Build GRCh38
rs766789167 Chr.19: 960101-960101 on Build GRCh38
rs1052153354 Chr.19: 960208-960208 on Build GRCh38
rs950860202 Chr.19: 960221-960221 on Build GRCh38
rs755197409 Chr.19: 960103-960103 on Build GRCh38
rs972773194 Chr.19: 960155-960155 on Build GRCh38
rs749528929 Chr.19: 960118-960118 on Build GRCh38
rs959701345 Chr.19: 960083-960083 on Build GRCh38
rs777850746 Chr.19: 960093-960093 on Build GRCh38
rs970653088 Chr.19: 960106-960106 on Build GRCh38
rs1031300636 Chr.19: 960077-960077 on Build GRCh38
rs770888541 Chr.19: 960046-960046 on Build GRCh38
rs1013905258 Chr.19: 960265-960265 on Build GRCh38

View Full Product Details