Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 173616229-173616474 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BOD1
Entrez Gene ID
91272
Gene Name
biorientation of chromosomes in cell division 1
Gene Aliases
FAM44B
UniGene
Hs.425091
Gene Chromosome Location
Chr.5: 173607145-173617077 on Build GRCh38
Transcripts
NM_001159651.1 NM_138369.2 XM_017010017.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5356579 c.94G>A p.A32T Chr.5: 173616343-173616343 on Build GRCh38
6209201 c.26G>A p.G9E Chr.5: 173616411-173616411 on Build GRCh38
6425459 c.146C>A p.P49Q Chr.5: 173616291-173616291 on Build GRCh38
5356578 c.94G>A p.A32T Chr.5: 173616343-173616343 on Build GRCh38
6612239 c.21C>G p.G7G Chr.5: 173616416-173616416 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773693940 Chr.5: 173616433-173616433 on Build GRCh38
rs745689308 Chr.5: 173616424-173616424 on Build GRCh38
rs761974596 Chr.5: 173616342-173616342 on Build GRCh38
rs780925841 Chr.5: 173616381-173616381 on Build GRCh38
rs749298438 Chr.5: 173616401-173616401 on Build GRCh38
rs749027913 Chr.5: 173616305-173616305 on Build GRCh38
rs780612809 Chr.5: 173616257-173616257 on Build GRCh38
rs769090853 Chr.5: 173616414-173616416 on Build GRCh38
rs774874725 Chr.5: 173616423-173616429 on Build GRCh38
rs770919204 Chr.5: 173616312-173616312 on Build GRCh38
rs768751082 Chr.5: 173616314-173616314 on Build GRCh38
rs914105610 Chr.5: 173616303-173616303 on Build GRCh38
rs769982410 Chr.5: 173616432-173616432 on Build GRCh38
rs868613881 Chr.5: 173616268-173616268 on Build GRCh38
rs938039578 Chr.5: 173616315-173616315 on Build GRCh38
rs528478868 Chr.5: 173616343-173616343 on Build GRCh38
rs763497502 Chr.5: 173616443-173616443 on Build GRCh38
rs11554340 Chr.5: 173616425-173616425 on Build GRCh38
rs766785149 Chr.5: 173616360-173616360 on Build GRCh38
rs548238064 Chr.5: 173616348-173616348 on Build GRCh38
rs866357017 Chr.5: 173616250-173616250 on Build GRCh38
rs752095035 Chr.5: 173616383-173616383 on Build GRCh38
rs962721906 Chr.5: 173616410-173616411 on Build GRCh38
rs886944241 Chr.5: 173616413-173616413 on Build GRCh38
rs757098185 Chr.5: 173616402-173616402 on Build GRCh38
rs61742044 Chr.5: 173616368-173616368 on Build GRCh38
rs1015919841 Chr.5: 173616410-173616410 on Build GRCh38
rs776645383 Chr.5: 173616301-173616301 on Build GRCh38
rs778893368 Chr.5: 173616412-173616412 on Build GRCh38
rs761029131 Chr.5: 173616313-173616313 on Build GRCh38
rs777570840 Chr.5: 173616394-173616394 on Build GRCh38
rs995414019 Chr.5: 173616367-173616367 on Build GRCh38
rs768712520 Chr.5: 173616426-173616426 on Build GRCh38
rs780349169 Chr.5: 173616383-173616406 on Build GRCh38
rs763297939 Chr.5: 173616349-173616349 on Build GRCh38
rs776534383 Chr.5: 173616318-173616318 on Build GRCh38
rs755657387 Chr.5: 173616389-173616389 on Build GRCh38
rs766695169 Chr.5: 173616447-173616447 on Build GRCh38
rs754468117 Chr.5: 173616253-173616253 on Build GRCh38
rs748109580 Chr.5: 173616427-173616427 on Build GRCh38
rs866576052 Chr.5: 173616252-173616252 on Build GRCh38
rs751187603 Chr.5: 173616366-173616366 on Build GRCh38
rs748083915 Chr.5: 173616290-173616290 on Build GRCh38
rs951443018 Chr.5: 173616391-173616391 on Build GRCh38
rs749631448 Chr.5: 173616413-173616414 on Build GRCh38
rs866110188 Chr.5: 173616280-173616280 on Build GRCh38

View Full Product Details