Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64219690-64219959 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FERMT3
Entrez Gene ID
83706
Gene Name
fermitin family member 3
Gene Aliases
KIND3, MIG-2, MIG2B, UNC112C, URP2, URP2SF
UniGene
Hs.180535
Gene Chromosome Location
Chr.11: 64206665-64223891 on Build GRCh38
Transcripts
NM_031471.5 NM_178443.2 XM_011545294.2 XM_011545295.2 XM_017018398.1 XM_017018399.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6344231 c.1034G>T p.S345I Chr.11: 64219744-64219744 on Build GRCh38
4684362 c.1079G>A p.R360Q Chr.11: 64219789-64219789 on Build GRCh38
4684364 c.1080-5C>T p.? Chr.11: 64219886-64219886 on Build GRCh38
3416092 c.1121G>A p.R374H Chr.11: 64219920-64219920 on Build GRCh38
4521903 c.1135G>A p.V379M Chr.11: 64219934-64219934 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761480844 Chr.11: 64219710-64219710 on Build GRCh38
rs761647934 Chr.11: 64219715-64219715 on Build GRCh38
rs368225468 Chr.11: 64219731-64219731 on Build GRCh38
rs140579293 Chr.11: 64219923-64219923 on Build GRCh38
rs771851319 Chr.11: 64219869-64219869 on Build GRCh38
rs372050214 Chr.11: 64219840-64219840 on Build GRCh38
rs764978388 Chr.11: 64219887-64219887 on Build GRCh38
rs779076021 Chr.11: 64219757-64219757 on Build GRCh38
rs748199007 Chr.11: 64219771-64219771 on Build GRCh38
rs781707563 Chr.11: 64219935-64219935 on Build GRCh38
rs746310738 Chr.11: 64219936-64219936 on Build GRCh38
rs369541420 Chr.11: 64219860-64219860 on Build GRCh38
rs1027548745 Chr.11: 64219882-64219882 on Build GRCh38
rs765425998 Chr.11: 64219827-64219827 on Build GRCh38
rs777640353 Chr.11: 64219848-64219849 on Build GRCh38
rs568273430 Chr.11: 64219797-64219797 on Build GRCh38
rs752494649 Chr.11: 64219894-64219894 on Build GRCh38
rs11231726 Chr.11: 64219867-64219867 on Build GRCh38
rs747475656 Chr.11: 64219787-64219787 on Build GRCh38
rs201564837 Chr.11: 64219709-64219709 on Build GRCh38
rs777854608 Chr.11: 64219784-64219784 on Build GRCh38
rs757320764 Chr.11: 64219924-64219924 on Build GRCh38
rs143722885 Chr.11: 64219886-64219886 on Build GRCh38
rs772191175 Chr.11: 64219774-64219774 on Build GRCh38
rs199626623 Chr.11: 64219850-64219850 on Build GRCh38
rs371355501 Chr.11: 64219861-64219861 on Build GRCh38
rs772852769 Chr.11: 64219737-64219737 on Build GRCh38
rs752870494 Chr.11: 64219832-64219832 on Build GRCh38
rs113773254 Chr.11: 64219828-64219828 on Build GRCh38
rs760590755 Chr.11: 64219811-64219811 on Build GRCh38
rs758652600 Chr.11: 64219833-64219833 on Build GRCh38
rs897637418 Chr.11: 64219914-64219914 on Build GRCh38
rs753575419 Chr.11: 64219733-64219733 on Build GRCh38
rs772877647 Chr.11: 64219810-64219810 on Build GRCh38
rs766237163 Chr.11: 64219814-64219814 on Build GRCh38
rs772785957 Chr.11: 64219872-64219872 on Build GRCh38
rs767084171 Chr.11: 64219809-64219809 on Build GRCh38
rs868278853 Chr.11: 64219806-64219806 on Build GRCh38
rs764357988 Chr.11: 64219921-64219921 on Build GRCh38
rs554245597 Chr.11: 64219799-64219799 on Build GRCh38
rs746211685 Chr.11: 64219859-64219859 on Build GRCh38
rs374311779 Chr.11: 64219874-64219874 on Build GRCh38
rs766312180 Chr.11: 64219875-64219875 on Build GRCh38
rs575353509 Chr.11: 64219717-64219717 on Build GRCh38
rs200818339 Chr.11: 64219842-64219842 on Build GRCh38
rs913856397 Chr.11: 64219711-64219711 on Build GRCh38
rs776484766 Chr.11: 64219879-64219879 on Build GRCh38
rs771347436 Chr.11: 64219788-64219788 on Build GRCh38
rs774186296 Chr.11: 64219800-64219800 on Build GRCh38
rs754773551 Chr.11: 64219819-64219819 on Build GRCh38
rs746954198 Chr.11: 64219849-64219857 on Build GRCh38
rs372151180 Chr.11: 64219723-64219723 on Build GRCh38
rs753634777 Chr.11: 64219933-64219933 on Build GRCh38
rs867531303 Chr.11: 64219919-64219919 on Build GRCh38
rs753667231 Chr.11: 64219818-64219818 on Build GRCh38
rs146822669 Chr.11: 64219895-64219895 on Build GRCh38
rs77762249 Chr.11: 64219728-64219728 on Build GRCh38
rs776996237 Chr.11: 64219795-64219795 on Build GRCh38
rs777764659 Chr.11: 64219841-64219841 on Build GRCh38

View Full Product Details