Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30108178-30108409 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDPD3
Entrez Gene ID
79153
Gene Name
glycerophosphodiester phosphodiesterase domain containing 3
Gene Aliases
GDE7
UniGene
Hs.289015
Gene Chromosome Location
Chr.16: 30104810-30113557 on Build GRCh38
Transcripts
NM_024307.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5948718 c.787C>T p.L263L Chr.16: 30108245-30108245 on Build GRCh38
311431 c.808C>T p.R270* Chr.16: 30108224-30108224 on Build GRCh38
3508728 c.762G>A p.S254S Chr.16: 30108378-30108378 on Build GRCh38
5021883 c.819+9G>T p.? Chr.16: 30108204-30108204 on Build GRCh38
4060081 c.779G>A p.R260K Chr.16: 30108253-30108253 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749264033 Chr.16: 30108292-30108294 on Build GRCh38
rs750269908 Chr.16: 30108364-30108364 on Build GRCh38
rs145295505 Chr.16: 30108378-30108378 on Build GRCh38
rs773761647 Chr.16: 30108276-30108276 on Build GRCh38
rs774652381 Chr.16: 30108288-30108288 on Build GRCh38
rs187205508 Chr.16: 30108263-30108263 on Build GRCh38
rs370294642 Chr.16: 30108268-30108268 on Build GRCh38
rs369610999 Chr.16: 30108289-30108289 on Build GRCh38
rs779749501 Chr.16: 30108370-30108370 on Build GRCh38
rs745351218 Chr.16: 30108200-30108200 on Build GRCh38
rs61764623 Chr.16: 30108295-30108295 on Build GRCh38
rs752854385 Chr.16: 30108238-30108238 on Build GRCh38
rs758398528 Chr.16: 30108365-30108365 on Build GRCh38
rs372587383 Chr.16: 30108346-30108346 on Build GRCh38
rs150037677 Chr.16: 30108212-30108212 on Build GRCh38
rs539393239 Chr.16: 30108330-30108330 on Build GRCh38
rs752774343 Chr.16: 30108323-30108323 on Build GRCh38
rs8061772 Chr.16: 30108204-30108204 on Build GRCh38
rs756311411 Chr.16: 30108239-30108239 on Build GRCh38
rs767729270 Chr.16: 30108234-30108234 on Build GRCh38
rs763555664 Chr.16: 30108217-30108217 on Build GRCh38
rs981660383 Chr.16: 30108237-30108237 on Build GRCh38
rs868560344 Chr.16: 30108259-30108259 on Build GRCh38
rs201362291 Chr.16: 30108283-30108283 on Build GRCh38
rs866548784 Chr.16: 30108226-30108226 on Build GRCh38
rs771497172 Chr.16: 30108284-30108284 on Build GRCh38
rs775245754 Chr.16: 30108205-30108205 on Build GRCh38
rs775295434 Chr.16: 30108269-30108269 on Build GRCh38
rs778669550 Chr.16: 30108257-30108257 on Build GRCh38
rs146701553 Chr.16: 30108383-30108383 on Build GRCh38
rs757270367 Chr.16: 30108256-30108256 on Build GRCh38
rs773706730 Chr.16: 30108215-30108215 on Build GRCh38
rs377072266 Chr.16: 30108345-30108345 on Build GRCh38
rs754725154 Chr.16: 30108326-30108326 on Build GRCh38
rs766771553 Chr.16: 30108223-30108223 on Build GRCh38
rs746718095 Chr.16: 30108273-30108273 on Build GRCh38
rs200607779 Chr.16: 30108343-30108343 on Build GRCh38
rs768390906 Chr.16: 30108274-30108274 on Build GRCh38
rs780666211 Chr.16: 30108379-30108379 on Build GRCh38
rs746495506 Chr.16: 30108374-30108374 on Build GRCh38
rs777505464 Chr.16: 30108291-30108292 on Build GRCh38
rs770584843 Chr.16: 30108335-30108336 on Build GRCh38
rs762273933 Chr.16: 30108210-30108210 on Build GRCh38
rs200117011 Chr.16: 30108224-30108224 on Build GRCh38
rs750124469 Chr.16: 30108196-30108196 on Build GRCh38
rs930094251 Chr.16: 30108298-30108298 on Build GRCh38
rs778917138 Chr.16: 30108362-30108362 on Build GRCh38
rs749664859 Chr.16: 30108380-30108380 on Build GRCh38
rs749009238 Chr.16: 30108241-30108241 on Build GRCh38

View Full Product Details