Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 10876376-10876633 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCM2
Entrez Gene ID
9247
Gene Name
glial cells missing homolog 2
Gene Aliases
GCMB, hGCMb
UniGene
Hs.227098
Gene Chromosome Location
Chr.6: 10873223-10881865 on Build GRCh38
Transcripts
NM_004752.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3857332 c.443C>T p.A148V Chr.6: 10876458-10876458 on Build GRCh38
6867242 c.449T>C p.F150S Chr.6: 10876452-10876452 on Build GRCh38
3857333 c.441C>T p.N147N Chr.6: 10876460-10876460 on Build GRCh38
461926 c.368C>A p.S123Y Chr.6: 10876533-10876533 on Build GRCh38
4427972 c.447C>A p.I149I Chr.6: 10876454-10876454 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1028183098 Chr.6: 10876468-10876468 on Build GRCh38
rs35786951 Chr.6: 10876551-10876551 on Build GRCh38
rs779456082 Chr.6: 10876540-10876540 on Build GRCh38
rs775445560 Chr.6: 10876489-10876489 on Build GRCh38
rs749006010 Chr.6: 10876562-10876562 on Build GRCh38
rs768698947 Chr.6: 10876583-10876583 on Build GRCh38
rs145276001 Chr.6: 10876499-10876499 on Build GRCh38
rs374228069 Chr.6: 10876565-10876565 on Build GRCh38
rs371217595 Chr.6: 10876593-10876593 on Build GRCh38
rs745649479 Chr.6: 10876471-10876471 on Build GRCh38
rs772327357 Chr.6: 10876608-10876608 on Build GRCh38
rs761951360 Chr.6: 10876600-10876600 on Build GRCh38
rs749188435 Chr.6: 10876580-10876580 on Build GRCh38
rs756502589 Chr.6: 10876420-10876420 on Build GRCh38
rs375087764 Chr.6: 10876458-10876458 on Build GRCh38
rs750802127 Chr.6: 10876417-10876417 on Build GRCh38
rs762040812 Chr.6: 10876505-10876505 on Build GRCh38
rs368814748 Chr.6: 10876459-10876459 on Build GRCh38
rs367671760 Chr.6: 10876416-10876416 on Build GRCh38
rs776335364 Chr.6: 10876462-10876462 on Build GRCh38
rs1040019354 Chr.6: 10876556-10876556 on Build GRCh38
rs779715308 Chr.6: 10876575-10876575 on Build GRCh38
rs766685638 Chr.6: 10876516-10876516 on Build GRCh38
rs927455956 Chr.6: 10876610-10876610 on Build GRCh38
rs146812948 Chr.6: 10876498-10876498 on Build GRCh38
rs780578785 Chr.6: 10876424-10876424 on Build GRCh38
rs756803427 Chr.6: 10876552-10876552 on Build GRCh38
rs761079176 Chr.6: 10876515-10876515 on Build GRCh38
rs759190203 Chr.6: 10876442-10876443 on Build GRCh38
rs767846876 Chr.6: 10876510-10876510 on Build GRCh38
rs931100023 Chr.6: 10876597-10876597 on Build GRCh38
rs930824118 Chr.6: 10876474-10876474 on Build GRCh38
rs769530379 Chr.6: 10876487-10876487 on Build GRCh38
rs749782575 Chr.6: 10876446-10876446 on Build GRCh38
rs754305249 Chr.6: 10876536-10876536 on Build GRCh38
rs377314144 Chr.6: 10876569-10876569 on Build GRCh38
rs542569618 Chr.6: 10876457-10876457 on Build GRCh38
rs373057036 Chr.6: 10876490-10876490 on Build GRCh38
rs755491340 Chr.6: 10876537-10876537 on Build GRCh38
rs184620420 Chr.6: 10876564-10876564 on Build GRCh38

View Full Product Details