Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 66934148-66934387 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM96B
Entrez Gene ID
51647
Gene Name
family with sequence similarity 96 member B
Gene Aliases
CGI-128, MIP18
UniGene
Hs.9825
Gene Chromosome Location
Chr.16: 66932055-66934423 on Build GRCh38
Transcripts
NM_016062.3 NR_024525.2 NR_046109.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4683313 c.6A>G p.V2V Chr.16: 66934359-66934359 on Build GRCh38
1609498 c.10G>A p.G4S Chr.16: 66934355-66934355 on Build GRCh38
5660837 c.61C>T p.Q21* Chr.16: 66934304-66934304 on Build GRCh38
7075663 c.39G>C p.E13D Chr.16: 66934326-66934326 on Build GRCh38
4407557 c.20T>G p.V7G Chr.16: 66934345-66934345 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747220172 Chr.16: 66934355-66934355 on Build GRCh38
rs757950623 Chr.16: 66934332-66934332 on Build GRCh38
rs753507526 Chr.16: 66934315-66934315 on Build GRCh38
rs1032084435 Chr.16: 66934172-66934172 on Build GRCh38
rs375950439 Chr.16: 66934365-66934365 on Build GRCh38
rs780408191 Chr.16: 66934362-66934362 on Build GRCh38
rs762975687 Chr.16: 66934280-66934280 on Build GRCh38
rs777761203 Chr.16: 66934262-66934262 on Build GRCh38
rs1021506487 Chr.16: 66934190-66934190 on Build GRCh38
rs763568182 Chr.16: 66934336-66934347 on Build GRCh38
rs202164467 Chr.16: 66934248-66934248 on Build GRCh38
rs756804609 Chr.16: 66934344-66934344 on Build GRCh38
rs778913077 Chr.16: 66934223-66934223 on Build GRCh38
rs554566674 Chr.16: 66934177-66934177 on Build GRCh38
rs1002905238 Chr.16: 66934168-66934168 on Build GRCh38
rs377034609 Chr.16: 66934209-66934209 on Build GRCh38
rs770135318 Chr.16: 66934267-66934267 on Build GRCh38
rs77470938 Chr.16: 66934220-66934220 on Build GRCh38
rs746181958 Chr.16: 66934352-66934352 on Build GRCh38
rs749856523 Chr.16: 66934364-66934364 on Build GRCh38
rs777492408 Chr.16: 66934334-66934334 on Build GRCh38
rs758003722 Chr.16: 66934236-66934236 on Build GRCh38
rs769960116 Chr.16: 66934171-66934172 on Build GRCh38
rs746259066 Chr.16: 66934259-66934259 on Build GRCh38
rs777165677 Chr.16: 66934186-66934186 on Build GRCh38
rs781470049 Chr.16: 66934252-66934252 on Build GRCh38
rs766789584 Chr.16: 66934345-66934346 on Build GRCh38
rs374829700 Chr.16: 66934368-66934368 on Build GRCh38
rs754812319 Chr.16: 66934218-66934218 on Build GRCh38
rs866384549 Chr.16: 66934321-66934321 on Build GRCh38
rs746699790 Chr.16: 66934251-66934251 on Build GRCh38
rs760029796 Chr.16: 66934197-66934197 on Build GRCh38
rs373802770 Chr.16: 66934202-66934202 on Build GRCh38
rs773140266 Chr.16: 66934335-66934336 on Build GRCh38
rs768758866 Chr.16: 66934289-66934289 on Build GRCh38
rs1047654980 Chr.16: 66934170-66934170 on Build GRCh38
rs752730258 Chr.16: 66934329-66934329 on Build GRCh38
rs11556909 Chr.16: 66934314-66934314 on Build GRCh38
rs200532434 Chr.16: 66934176-66934176 on Build GRCh38
rs372085708 Chr.16: 66934173-66934173 on Build GRCh38
rs751244375 Chr.16: 66934337-66934337 on Build GRCh38
rs774353687 Chr.16: 66934366-66934366 on Build GRCh38
rs778342526 Chr.16: 66934174-66934174 on Build GRCh38
rs1013865591 Chr.16: 66934331-66934331 on Build GRCh38
rs780903835 Chr.16: 66934346-66934346 on Build GRCh38
rs747329200 Chr.16: 66934180-66934180 on Build GRCh38
rs762076560 Chr.16: 66934300-66934300 on Build GRCh38
rs776099590 Chr.16: 66934277-66934277 on Build GRCh38
rs760426982 Chr.16: 66934208-66934208 on Build GRCh38
rs764264118 Chr.16: 66934204-66934204 on Build GRCh38
rs866700118 Chr.16: 66934349-66934349 on Build GRCh38
rs751807113 Chr.16: 66934205-66934205 on Build GRCh38
rs1035279671 Chr.16: 66934272-66934272 on Build GRCh38
rs1044259546 Chr.16: 66934215-66934215 on Build GRCh38
rs369698544 Chr.16: 66934214-66934214 on Build GRCh38
rs950590013 Chr.16: 66934350-66934350 on Build GRCh38
rs907529466 Chr.16: 66934249-66934249 on Build GRCh38
rs774549089 Chr.16: 66934292-66934292 on Build GRCh38
rs756526985 Chr.16: 66934360-66934360 on Build GRCh38
rs771472860 Chr.16: 66934181-66934181 on Build GRCh38
rs769906744 Chr.16: 66934201-66934201 on Build GRCh38
rs373271757 Chr.16: 66934227-66934227 on Build GRCh38
rs759405706 Chr.16: 66934317-66934317 on Build GRCh38
rs777545325 Chr.16: 66934246-66934246 on Build GRCh38

View Full Product Details